Skip to Content
Merck
All Photos(3)

Key Documents

214698

Sigma-Aldrich

Pyridinium dichromate

98%

Synonym(s):

PDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2 · H2Cr2O7
CAS Number:
Molecular Weight:
376.20
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reagent type: oxidant

mp

152-153 °C (lit.)

SMILES string

c1ccncc1.c2ccncc2.O[Cr](=O)(=O)O[Cr](O)(=O)=O

InChI

1S/2C5H5N.2Cr.2H2O.5O/c2*1-2-4-6-5-3-1;;;;;;;;;/h2*1-5H;;;2*1H2;;;;;/q;;2*+1;;;;;;;/p-2

InChI key

RCBVKBFIWMOMHF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Pyridinium dichromate is a pyridinium salt and strong oxidizing agent to convert primary alcohols and secondary alcohols to aldehydes and ketones respectively.

Application

Oxidizing agent for conversion of primary alcohols to aldehydes and ketones, acetals to esters, and didehydroketones to enones in the presence of tert-butyl hydroperoxide (cat. no. 213128).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Mol. Catal. A: Chem., 250, 70-70 (2006)
J M Bokhorst et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 33(5), 825-833 (2019-12-18)
Tumor budding is a promising and cost-effective biomarker with strong prognostic value in colorectal cancer. However, challenges related to interobserver variability persist. Such variability may be reduced by immunohistochemistry and computer-aided tumor bud selection. Development of computer algorithms for this
Sameer Shivji et al.
Histopathology, 77(3), 351-368 (2020-05-03)
Poorly differentiated clusters (PDC), defined as small groups of ≥5 tumour cells without glandular differentiation, have gained recent attention as a promising prognostic factor in colorectal cancer (CRC). Numerous studies have shown PDC to be significantly associated with other adverse
Synthetic Communications, 23, 2701-2701 (1993)
Kinetics and mechanism of oxidation of pimary alcohols by pyridinium dichromate
Harit H, et al.
Chemical Science, 4, 49-58 (2015)

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service