Skip to Content
Merck
All Photos(3)

Key Documents

190144

Sigma-Aldrich

Pyridinium chlorochromate

98%

Synonym(s):

PCC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H6NClCrO3
CAS Number:
Molecular Weight:
215.56
Beilstein:
7054643
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reagent type: oxidant

mp

205-208 °C (lit.)

SMILES string

O[Cr](Cl)(=O)=O.c1ccncc1

InChI

1S/C5H5N.ClH.Cr.H2O.2O/c1-2-4-6-5-3-1;;;;;/h1-5H;1H;;1H2;;/q;;+2;;;/p-2

InChI key

JWIPGAFCGUDKEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

PCC is an important organic reagent used for oxidation reactions.

Application

Stable versatile oxidizing agent for mild and selective oxidation of alcohols.
Used to oxidize an allylic methylene group in a synthesis of α,ß-unsaturated lactones from D-glucose using Grubbs-catalyzed ring-closing metathesis.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Ox. Sol. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xueling Zhu et al.
Scientific reports, 7, 43105-43105 (2017-02-23)
Neuroimaging evidence implicates the association between rumination and default mode network (DMN) in major depressive disorder (MDD). However, the relationship between rumination and DMN subsystems remains incompletely understood, especially in patients with MDD. Thirty-three first-episode drug-naive patients with MDD and
Synthesis, 245-245 (1982)
Pegah Khosravi et al.
Algorithms for molecular biology : AMB, 10, 23-23 (2015-07-15)
Knowledge of interaction types in biological networks is important for understanding the functional organization of the cell. Currently information-based approaches are widely used for inferring gene regulatory interactions from genomics data, such as gene expression profiles; however, these approaches do
Anna M Larsson et al.
Journal of experimental botany, 68(14), 3857-3867 (2017-04-04)
Cyanobacterial CO2 fixation is promoted by encapsulating and co-localizing the CO2-fixing enzymes within a protein shell, the carboxysome. A key feature of the carboxysome is its ability to control selectively the flux of metabolites in and out of the shell.
Tetrahedron, 62, 11165-11165 (2006)

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service