Skip to Content
Merck
All Photos(4)

Key Documents

130079

Sigma-Aldrich

Methyltriphenylphosphonium bromide

98%

Synonym(s):

TPMP, Triphenylmethylphosphonium bromide, triphenylmethylphosphonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3P(C6H5)3Br
CAS Number:
Molecular Weight:
357.22
Beilstein:
3599467
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

230-234 °C (lit.)

functional group

phosphine

SMILES string

[Br-].C[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C19H18P.BrH/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;/h2-16H,1H3;1H/q+1;/p-1

InChI key

LSEFCHWGJNHZNT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used widely for methylenation via the Wittig reaction
Wittig olefination salt employed recently in a synthesis of an enyne which, via ring closing metathesis, provided a benzazepine ring system.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

>464.0 °F

Flash Point(C)

> 240 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duncan G G McMillan et al.
The Journal of biological chemistry, 282(24), 17395-17404 (2007-04-17)
Analysis of the atp operon from the thermoalkaliphilic Bacillus sp. TA2.A1 and comparison with other atp operons from alkaliphilic bacteria reveals the presence of a conserved lysine residue at position 180 (Bacillus sp. TA2.A1 numbering) within the a subunit of
L Calzada et al.
Archives of andrology, 38(2), 151-155 (1997-03-01)
Membrane potential across the plasma membrane (psi) of human sperm was determined by measuring the accumulation of triphenylmethylphosphonium (TPMP+). Washed sperm of fertile or infertile men were suspended, incubated in low-K+ and high-Na+ or high-K+ and low-Na+ medium, and allowed
D P Nolan et al.
European journal of biochemistry, 267(15), 4615-4623 (2000-07-21)
The plasma-membrane potential (Delta(psi)p) in bloodstream forms of Trypanosoma brucei was studied using several different radiolabelled probes: 86Rb+ and [14C]SCN- were used to report Delta(psi)p directly because they distribute in easily measured quantities across the plasma membrane only, and [3H]methyltriphenylphosphonium
Geoffrey F Kelso et al.
Annals of the New York Academy of Sciences, 959, 263-274 (2002-04-27)
Mitochondrial-targeted antioxidants that selectively block mitochondrial oxidative damage and prevent some types of cell death have been developed. These antioxidants are ubiquinone and tocopherol derivatives and are targeted to mitochondria by covalent attachment to a lipophilic triphenylphosphonium cation. Because of
Meredith F Ross et al.
The Biochemical journal, 400(1), 199-208 (2006-09-05)
Lipophilic monocations can pass through phospholipid bilayers and accumulate in negatively-charged compartments such as the mitochondrial matrix, driven by the membrane potential. This property is used to visualize mitochondria, to deliver therapeutic molecules to mitochondria and to measure the membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service