Skip to Content
Merck
All Photos(3)

Key Documents

122564

Sigma-Aldrich

Mechlorethamine hydrochloride

98%

Synonym(s):

2-Chloro-N-(2-chloroethyl)-N-methylethanamine, N-Methylbis(2-chloroethyl)amine hydrochloride, Bis(2-Chloroethyl)methylamine, Bis(2-chloroethyl)methylamine, MBA hydrochloride, NM, Nitrogen mustard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCH2CH2)2NCH3·HCl
CAS Number:
Molecular Weight:
192.51
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

108-111 °C (lit.)

solubility

H2O: very soluble
alcohol: soluble

functional group

amine
chloro

SMILES string

Cl.CN(CCCl)CCCl

InChI

1S/C5H11Cl2N.ClH/c1-8(4-2-6)5-3-7;/h2-5H2,1H3;1H

InChI key

QZIQJVCYUQZDIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mechlorethamine hydrochloride was used in a fluorescence-based apurinic/apyrimidinic site cleavage assay for the identification of apurinic/apyrimidinic endonuclease inhibitors in human cells. It was used in a study to demonstrate the effect of povidone iodine ointment against skin toxicity caused by sulphur and nitrogen mustard.

Biochem/physiol Actions

Mechlorethamine hydrochloride administration reduces the tensile strength of the abdominal wounds in rats.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Carc. 1B - Eye Dam. 1 - Muta. 1B - Repr. 1B - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingcui Song et al.
Experimental cell research, 382(1), 111450-111450 (2019-06-04)
The metastasis-associated gene 1 (MTA1) has previously been recognized as an oncogene in many tumors, and aberrant MTA1 expression has been related to invasion and migration; however, its role and underlying molecular mechanism in oral squamous carcinoma (OSCC) remain largely
Aliete Wan et al.
Human mutation, 40(8), 1127-1144 (2019-04-13)
Characterizing the pathogenicity of DNA sequence variants of unknown significance (VUS) is a major bottleneck in human genetics, and is increasingly important in determining which patients with inherited retinal diseases could benefit from gene therapy. A library of 210 rhodopsin
Chao Wu et al.
General and comparative endocrinology, 281, 91-104 (2019-05-24)
Two structurally related peptides, arginine vasotocin (AVT) and mesotocin (MT), are reported to regulate many physiological processes, such as anti-diuresis and oviposition in birds, and their actions are likely mediated by four AVT/MT receptors (AVPR1A, AVPR1B, MTR and AVPR2b), which
Jin Zhang et al.
Journal of molecular and cellular cardiology, 134, 74-85 (2019-07-16)
Sick sinus syndrome (SSS) is primarily a disease of the elderly, and age-dependent decrease in Cav1.2 and Cav1.3 Ca2+ channels within the sinus node has been shown to play an important role in sinoatrial node (SAN) degeneration; however, posttranscriptional mechanisms
U Wormser et al.
Archives of toxicology, 71(3), 165-170 (1997-01-01)
Mustard gas (sulphur mustard, SM) is a powerful vesicant employed as a chemical weapon. The present study demonstrates the effect of povidone iodine (PI) ointment against skin toxicity caused by SM. Gross and histopathological examinations showed that application of PI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service