Skip to Content
Merck
All Photos(2)

Key Documents

417602

Sigma-Aldrich

2-Chloroethyl phenyl sulfide

98%

Synonym(s):

2-(Phenylthio)ethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2SC6H5
CAS Number:
Molecular Weight:
172.68
Beilstein:
1907116
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.583 (lit.)

bp

90-91 °C/1 mmHg (lit.)

density

1.174 g/mL at 25 °C (lit.)

SMILES string

ClCCSc1ccccc1

InChI

1S/C8H9ClS/c9-6-7-10-8-4-2-1-3-5-8/h1-5H,6-7H2

InChI key

QDXIHHOPZFCEAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloroethyl phenyl sulfide has been reported as sulfur mustard simulant. It is a chemical-warfare agent (CWA) simulant and its oxidation using hydrothane films has been reported. It is also reported as a simulant for the chemical warfare agent mustard [bis(2-chloroethyl) sulfide]. Its reaction with vesicular functionalized double-chain surfactant (potassium O, O′-didodecylphosphorodithioate) at pH 9.0 borate buffer affords [S-[(2-phenylthio)ethyl]O,O′-didodecylphosphorodithioate]. Its oxidation with H2O2 catalyzed by [MnL](ClO4)2 (L = N,N′,N″-tris(2-hydroxypropyl)-1,4,7-triazacyclononane) afforded 2-chloroethyl phenyl sulfone and phenyl vinyl sulfone.

Application

2-Chloroethyl phenyl sulfide, a sulfur mustard agent stimulant, may be used as target analyte in a bioassay for the detection of mustard agent.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Carc. 1A - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabiana Arduini et al.
Sensors (Basel, Switzerland), 15(2), 4353-4367 (2015-02-18)
In this work a novel bioassay for mustard agent detection was proposed. The bioassay is based on the capability of these compounds to inhibit the enzyme choline oxidase. The enzymatic activity, which is correlated to the mustard agents, was electrochemically
Raymond T Gephart et al.
ACS applied materials & interfaces, 5(20), 10191-10200 (2013-09-26)
Using reactive singlet oxygen (1O2), the oxidation of chemical-warfare agent (CWA) simulants has been demonstrated. The zinc octaphenoxyphthalocyanine (ZnOPPc) complex was demonstrated to be an efficient photosensitizer for converting molecular oxygen (O2) to 1O2 using broad-spectrum light (450-800 nm) from
Reaction of a vesicular functionalized surfactant with 2-chloroethyl phenyl sulfide, a mustard simulant.
Jaeger DA, et al.
Langmuir, 15(21), 7180-7185 (1999)
Sulfide oxidation with H2O2 catalyzed by manganese complex of N,N',N?-tris(2-hydroxypropyl)-1, 4, 7-triazacyclononane.
Feng X-M, et al.
Inorgorganica Chimica Acta, 360(15), 4103-4110 (2007)
Quaternary ammonium O, O'-dialkylphosphorodithioate surfactants and their reactions with 2-chloroethyl phenyl sulfide, a mustard simulant.
Jaeger DA, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 196(2), 209-216 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service