Merck
  • Functional characterization of two acyltransferases from Populus trichocarpa capable of synthesizing benzyl benzoate and salicyl benzoate, potential intermediates in salicinoid phenolic glycoside biosynthesis.
Functional characterization of two acyltransferases from Populus trichocarpa capable of synthesizing benzyl benzoate and salicyl benzoate, potential intermediates in salicinoid phenolic glycoside biosynthesis.
Phytochemistry (2015-01-07)
Russell J Chedgy, Tobias G Köllner, C Peter Constabel
ABSTRACT
Salicinoids are phenolic glycosides (PGs) characteristic of the Salicaceae and are known defenses against insect herbivory. Common examples are salicin, salicortin, tremuloidin, and tremulacin, which accumulate to high concentrations in the leaves and bark of willows and poplars. Although their biosynthetic pathway is not known, recent work has suggested that benzyl benzoate may be a potential biosynthetic intermediate. Two candidate genes, named PtACT47 and PtACT49, encoding BAHD-type acyl transferases were identified and are predicted to produce such benzylated secondary metabolites. Herein described are the cDNA cloning, heterologous expression and in vitro functional characterization of these two BAHD acyltransferases. Recombinant PtACT47 exhibited low substrate selectivity and could utilize acetyl-CoA, benzoyl-CoA, and cinnamoyl-CoA as acyl donors with a variety of alcohols as acyl acceptors. This enzyme showed the greatest Km/Kcat ratio (45.8 nM(-1) s(-1)) and lowest Km values (45.1 μM) with benzoyl-CoA and salicyl alcohol, and was named benzoyl-CoA: salicyl alcohol O-benzoyltransferase (PtSABT). Recombinant PtACT49 utilized a narrower range of substrates, including benzoyl-CoA and acetyl-CoA and a limited number of alcohols. Its highest Km/Kcat (31.8 nM(-1) s(-1)) and lowest Km (55.3 μM) were observed for benzoyl-CoA and benzyl alcohol, and it was named benzoyl-CoA: benzyl alcohol O-benzoyltransferase (PtBEBT). Both enzymes were also capable of synthesizing plant volatile alcohol esters, such as hexenyl benzoate, at trace levels. Although the activities demonstrated are consistent with roles in salicinoid biosynthesis, direct tests of this hypothesis using transgenic poplar must still be performed.
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
2-Hydroxybenzyl alcohol, 99%
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
2-Acetonaphthone, 99%
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Benzyl benzoate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Benzyl alcohol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, SAJ first grade, ≥98.5%
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Deuterium, 99.96 atom % D
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Cinnamyl alcohol, ≥98%, FG
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl benzoate, natural, ≥99%, FG
Sigma-Aldrich
Benzyl benzoate, ≥99%, FCC, FG
Sigma-Aldrich
Cinnamyl alcohol, 98%
Sigma-Aldrich
Benzyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Sigma-Aldrich
Methyl β-naphthyl ketone, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG