Skip to Content
Merck
All Photos(1)

Documents

03-1670

Sigma-Aldrich

Benzyl alcohol

SAJ first grade, ≥98.5%

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

Assay

≥98.5%

form

liquid

autoignition temp.

817 °F

availability

available only in Japan

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Béatrice Hechler et al.
The Journal of pharmacology and experimental therapeutics, 314(1), 232-243 (2005-03-29)
Our aim was to determine whether the newly described P2X1 antagonist NF449 [4,4',4'',4'''-(carbonylbis(imino-5,1,3-benzenetriylbis(carbonylimino)))tetrakis-benzene-1,3-disulfonic acid octasodium salt] could selectively antagonize the platelet P2X1 receptor and how it affected platelet function. NF449 inhibited alpha,beta-methyleneadenosine 5'-triphosphate-induced shape change (IC50 = 83 +/- 13
Yuma Morimoto et al.
Inorganic chemistry, 51(18), 10025-10036 (2012-09-08)
The rate of oxidation of 2,5-dimethoxybenzyl alcohol (2,5-(MeO)(2)C(6)H(3)CH(2)OH) by [Fe(IV)(O)(N4Py)](2+) (N4Py = N,N-bis(2-pyridylmethyl)-N-bis(2-pyridyl)methylamine) was enhanced significantly in the presence of Sc(OTf)(3) (OTf(-) = trifluoromethanesulfonate) in acetonitrile (e.g., 120-fold acceleration in the presence of Sc(3+)). Such a remarkable enhancement of the
A P De Leenheer et al.
Clinical chemistry, 31(1), 142-146 (1985-01-01)
For this sensitive RIA for 1 alpha,25-dihydroxyvitamin D, we used antibodies to 1 alpha,25-dihydroxycholecalciferol-3-hemisuccinate conjugated to bovine serum albumin, raised in rabbits. These antibodies show a high affinity for 1 alpha,25-dihydroxyvitamin D3 but cross react with other vitamin D metabolites
Qing Lan et al.
Science (New York, N.Y.), 306(5702), 1774-1776 (2004-12-04)
Benzene is known to have toxic effects on the blood and bone marrow, but its impact at levels below the U.S. occupational standard of 1 part per million (ppm) remains uncertain. In a study of 250 workers exposed to benzene
Meenakshisundaram Sankar et al.
ACS nano, 6(8), 6600-6613 (2012-07-10)
We report a convenient excess anion modification and post-reduction step to the impregnation method which permits the reproducible preparation of supported bimetallic AuPd nanoparticles having a tight particle size distribution comparable to that found for sol-immobilization materials but without the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service