Skip to Content
Merck
All Photos(1)

Key Documents

13-0850

Sigma-Aldrich

Hexane

≥96.5%, suitable for HPLC

Synonym(s):

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
41116023
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Product Name

Hexane, suitable for HPLC

grade

HPLC grade

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Assay

≥96.5%

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

availability

available only in Japan

technique(s)

HPLC: suitable

refractive index

n20/D 1.375 (lit.)

pH

7

bp

69 °C (lit.)

mp

−95 °C (lit.)

density

0.659 g/mL at 25 °C (lit.)

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Matti Marklund et al.
Circulation, 132(7), 586-594 (2015-06-19)
High intake of polyunsaturated fatty acids (PUFAs) may reduce the risk of cardiovascular disease (CVD) and mortality. Large, prospective studies including both sexes and circulating PUFAs as dietary biomarkers are needed. We investigated sex-specific associations of the major dietary PUFAs
Alexandre S Cristino et al.
Nature communications, 5, 5529-5529 (2014-11-21)
Increasing evidence suggests small non-coding RNAs (ncRNAs) such as microRNAs (miRNAs) control levels of mRNA expression during experience-related remodelling of the brain. Here we use an associative olfactory learning paradigm in the honeybee Apis mellifera to examine gene expression changes
Hilal E Toraman et al.
Journal of chromatography. A, 1359, 237-246 (2014-07-30)
The detailed compositional characterization of plastic waste pyrolysis oil was performed with comprehensive two-dimensional GC (GC×GC) coupled to four different detectors: a flame ionization detector (FID), a sulfur chemiluminescence detector (SCD), a nitrogen chemiluminescence detector (NCD) and a time of
Ioannis Lignos et al.
Small (Weinheim an der Bergstrasse, Germany), 11(32), 4009-4017 (2015-05-23)
The early-time kinetics (<1 s) of lead sulfide (PbS) quantum dot formation are probed using a novel droplet-based microfluidic platform, which allows for high-throughput and real-time optical analysis of the reactive process with millisecond time resolution. The reaction platform enables
R Lebrero et al.
Water research, 47(7), 2199-2212 (2013-03-19)
The performance of a flat-membrane biofilm reactor (MBR) for the removal of acetone, toluene, limonene and hexane at concentrations ranging from 1.3 to 3.2 mg m(-3) was investigated at different gas residence times (GRT): 60, 30, 15 and 7 s. A preliminary abiotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service