Skip to Content
Merck
All Photos(1)

Documents

166952

Sigma-Aldrich

2-Hydroxybenzyl alcohol

99%

Synonym(s):

Salicyl alcohol, Saligenin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2OH
CAS Number:
Molecular Weight:
124.14
Beilstein:
1907195
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

83-85 °C (lit.)

solubility

ethanol: soluble 5%, clear to very slightly hazy, colorless to light yellow

SMILES string

OCc1ccccc1O

InChI

1S/C7H8O2/c8-5-6-3-1-2-4-7(6)9/h1-4,8-9H,5H2

InChI key

CQRYARSYNCAZFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxybenzyl alcohol participates in the selective ring opening reaction of 4H-1,3,2-benzodioxasilines.

2-Hydroxybenzyl alcohol can be used as a coupling reagent to synthesize O-heterocycles.

Application

2-Hydroxybenzyl alcohol was used in gastrodin production via biotransformation by cultured cells of Aspergillus foetidus and Penicillium cyclopium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linlin Fan et al.
Applied biochemistry and biotechnology, 170(1), 138-148 (2013-03-14)
The objective of this work was to take advantage of the resting cells of suitable fungus as an in vitro model to prepare gastrodin from p-2-hydroxybenzyl alcohol (HBA), which mainly exists in the metabolites of the plant Gastrodia elata Blume.
Fernando A Genta et al.
Journal of insect physiology, 52(6), 593-601 (2006-04-08)
Tenebrio molitor larvae were successfully reared free of cultivatable gut lumen bacteria, yeasts and fungi using two approaches; aseptic rearing from surface sterilized eggs and by feeding larvae with antibiotic-containing food. Insects were reared on a rich-nutrient complete diet or
Teruaki Akao et al.
Planta medica, 68(8), 714-718 (2002-09-11)
Pharmacokinetic and pharmacological studies were performed to compare the antipyretic effects of salicin (SL), saligenin (SG, an aglycone of SL) and salicylic acid (SA, an active metabolite of SL) in rats. When SL was administered orally to rats, SA appeared
Nicolas Gisch et al.
Journal of medicinal chemistry, 51(21), 6752-6760 (2008-10-07)
Recently we reported on conceptually new enzymatically activated cycloSal-pronucleotides. Now, we developed this concept further with new compounds of this type. The basic idea is fast intracellular cleavage of a functionalized group at the cycloSal residue that results in a
René Roberto Bodemann et al.
Proceedings. Biological sciences, 279(1745), 4126-4134 (2012-08-10)
Allomones are widely used by insects to impede predation. Frequently these chemical stimuli are released from specialized glands. The larvae of Chrysomelina leaf beetles produce allomones in gland reservoirs into which the required precursors and also the enzymes are secreted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service