Merck
  • Stable chlorine isotope analysis of chlorinated acetic acids using gas chromatography/quadrupole mass spectrometry.
Stable chlorine isotope analysis of chlorinated acetic acids using gas chromatography/quadrupole mass spectrometry.
Rapid communications in mass spectrometry : RCM (2015-11-14)
Milena E Miska, Orfan Shouakar-Stash, Henry Holmstrand
ABSTRACT
The environmental occurrence of chlorinated acetic acids (CAAs) has been extensively studied, but the sources and transport are still not yet fully understood. A promising approach for source apportionment and process studies is the isotopic characterization of target compounds. We present the first on-line stable chlorine isotope analysis of CAAs by use of gas chromatography/quadrupole mass spectrometry (GC/qMS). Following approved procedures for concentration analysis, CAAs extracted into MTBE were methylated to GC-amenable methyl esters (mCAAs). These mCAAs were then analyzed by GC/qMS for their stable chlorine isotope composition using a sample/standard-bracketing approach (CAA standards in the range δ(37) Cl -6.3 to -0.2 ‰, Standard Mean Ocean Chloride). Cross-calibration of the herein presented method with off-line reference methods (thermal ionization and continuous-flow GC isotope ratio mass spectrometry; TI-MS and CF-GC/IRMS, respectively) shows good agreement between the methods (regression slope for GC/qMS vs reference method data sets: 0.92 ± 0.29). Sample amounts as small as 10 pmol Cl can herewith be analyzed with a precision of 0.1 to 0.4 ‰. This method should be useful for environmental studies of CAAs at ambient concentrations in precipitations (<0.06 to 100 nmol L(-1) ), surface waters (<0.2 to 5 nmol L(-1) ) and soil (<0.6 to 2000 nmol kg(-1) dry soil) where conventional off-line methods cannot be applied.
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
DCA Deblock (0.36M dichloroacetic acid in toluene)
Sigma-Aldrich
Copper(I) chloride, JIS special grade, ≥95.0%
Sigma-Aldrich
Chloroacetic acid, JIS special grade, ≥99.0%
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Chlorine, ≥99.5%
Sigma-Aldrich
Iodomethane-12C, ≥99.9 atom % 12C, ≥99% (CP), contains copper as stabilizer
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Chloroacetic acid, ACS reagent, ≥99.0%
Supelco
tert-Butyl methyl ether, HPLC grade, suitable for HPLC, 99.8%
Sigma-Aldrich
Chloroacetic acid, 99%
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Iodomethane, SAJ special grade, ≥99.5%
Sigma-Aldrich
tert-Butyl methyl ether, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Copper(I) chloride, ReagentPlus®, purified, ≥99%
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%