Skip to Content
Merck
All Photos(2)

Key Documents

224332

Sigma-Aldrich

Copper(I) chloride

ReagentPlus®, purified, ≥99%

Synonym(s):

Copper monochloride, Cuprous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl
CAS Number:
Molecular Weight:
99.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
form:
powder
Pricing and availability is not currently available.

vapor pressure

1.3 mmHg ( 546 °C)

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

powder

reaction suitability

reagent type: catalyst
core: copper

pH

5 (20 °C, 50 g/L)

bp

1490 °C (lit.)

mp

430 °C (lit.)

SMILES string

Cl[Cu]

InChI

1S/ClH.Cu/h1H;/q;+1/p-1

InChI key

OXBLHERUFWYNTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cu is present in +1 (cuprous) valence state in CuCl.[1] CuCl is a semiconductor having a wide band gap. It is a promising candidate for use in silicon-based optoelectronics since its lattice resembles with silicon. It participates in various reactions with Grignard reagents.[2]

Application

Copper(I) chloride (CuCl) has been used in the synthesis of copper nanoring structures.[3] It has also been used as a precursor for the fabrication of ordered array of TiO2 nanoparticles on the conductive nitrogen-doped graphene (NG).[4]
Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper (I) Chloride.
Bertz SH, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2007)
Network of Heterogeneous Catalyst Arrays on the Nitrogen-Doped Graphene for Synergistic Solar Energy Harvesting of Hydrogen from Water.
Shin, Sang Rim, et al.
Chemistry of Materials (2016)
Solvothermal synthesis and mechanical characterization of single crystalline copper nanorings.
Zhan Y, et al.
Journal of Crystal Growth, 325(1), 76-80 (2011)
Shuxin Wang et al.
Nature communications, 8(1), 848-848 (2017-10-12)
It has long been a challenge to dope metal nanoparticles with a specific number of heterometal atoms at specific positions. This becomes even more challenging if the heterometal belongs to the same group as the host metal because of the
Erwan Gicquel et al.
Biomacromolecules, 20(7), 2545-2556 (2019-06-28)
This study investigates the adsorption of a block copolymer composed of a poly(2-(dimethylamino)ethyl methacrylate) (PDMAEMA) cationic polyelectrolyte and a poly(di(ethylene glycol) methyl ethermethacrylate) (PDEGMA) on oxidized cellulose nanocrystals (TO-CNCs) to produce hydrogels. PDMEAMA- b-PDEGMA was synthesized by atom-transfer radical polymerization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service