Merck
  • Novel imidazolium-embedded N,N-dimethylaminopropyl-functionalized silica-based stationary phase for hydrophilic interaction/reversed-phase mixed-mode chromatography.
Novel imidazolium-embedded N,N-dimethylaminopropyl-functionalized silica-based stationary phase for hydrophilic interaction/reversed-phase mixed-mode chromatography.
Analytical and bioanalytical chemistry (2015-10-03)
Shijia Liu, Hongxin Xu, Jiaojiao Yu, Danyang Li, Mingyan Li, Xiaoqiang Qiao, Xinying Qin, Hongyuan Yan
ABSTRACT
A novel imidazolium-embedded N,N-dimethylaminopropyl-functionalized silica-based stationary phase (Sil-ImCl) was prepared and further used for hydrophilic interaction/reversed-phase mixed-mode chromatography. The Sil-ImCl stationary phase was respectively characterized by Fourier transform infrared spectrometry, thermogravimetric analysis, and element analysis. A variety of hydrophilic or hydrophobic compounds were used to evaluate the retention mechanisms of the developed stationary phase, and the effects of buffer salt concentration and pH of mobile phase on the retention of these compounds were also investigated. The developed stationary phase was successfully applied for separation of nucleosides and nucleic acid bases, water-soluble vitamins, phenols, and positional isomers. Moreover, simultaneous separation of polar and nonpolar compounds was also achieved with high resolution, outperforming the commercially available C8 column and amino column. Furthermore, the Sil-ImCl stationary phase has been successfully applied for separation of secondary metabolites of Hansfordia sinuosae. All these results demonstrate that the Sil-ImCl stationary phase might be promising for separation of complex polar and nonpolar compounds with high efficiency, especially in biological industry.
Sigma-Aldrich
(3-Mercaptopropyl)trimethoxysilane, 95%
Sigma-Aldrich
Triphenylene, 98%
Sigma-Aldrich
Inosine, ≥99% (HPLC)
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
o-Terphenyl, 99%
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
1-Vinylimidazole, ≥99%
Sigma-Aldrich
m-Terphenyl, 99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), SAJ first grade, ≥98.0%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%