Skip to Content
Merck
All Photos(1)

Key Documents

T82600

Sigma-Aldrich

Triphenylene

98%

Synonym(s):

9,10-Benzophenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1342908
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

438 °C (lit.)

mp

195-198 °C (lit.)

fluorescence

λex 265 nm; λem 372 nm in cyclohexane

SMILES string

c1ccc2c(c1)c3ccccc3c4ccccc24

InChI

1S/C18H12/c1-2-8-14-13(7-1)15-9-3-4-11-17(15)18-12-6-5-10-16(14)18/h1-12H

InChI key

SLGBZMMZGDRARJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Md Akhtarul Alam et al.
Journal of the American Chemical Society, 131(49), 17722-17723 (2009-12-10)
Triphenylene (TP) derivatives bearing appropriate paraffinic side chains with imidazolium ion-based ionic liquid (IL) pendants were unveiled to display a phase diagram with liquid crystalline (LC) mesophases of bicontinuous cubic (Cub(bi)) and hexagonal columnar (Col(h)) geometries. While their phase transition
Martin Kaller et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(21), 6326-6337 (2010-04-23)
The twisted lateral tetraalkyloxy ortho-terphenyl units in dibenzo[18]crown-6 ethers 1a-f were readily converted into the flat tetraalkyloxytriphenylene systems 2a-f by oxidative cyclization with FeCl(3) in nitromethane. Reactions of the latter with potassium salts gave complexes KX.2, which displayed mesomorphic properties.
Lucas A Haverkate et al.
The journal of physical chemistry. B, 116(43), 13098-13105 (2012-10-05)
Discotic liquid crystalline (DLC) charge transfer (CT) complexes, which combine visible light absorption with rapid charge transfer characteristics within the CT complex, can have a great potential for photovoltaic applications when they can be made to self-assemble in a bulk
Vandana Bhalla et al.
Dalton transactions (Cambridge, England : 2003), 41(37), 11413-11418 (2012-08-15)
The binding behavior of triphenylene based copper ensemble prepared in situ has been investigated toward various anions (F(-), Cl(-), Br(-), I(-), CH(3)COO(-), H(2)PO(4)(-), NO(3)(-), OH(-), ClO(4)(-), CN(-), CO(3)(-) and SO(4)(-)) by UV-vis and fluorescence spectroscopy. Among various anions tested, 1-Cu(2+)
Ozcan Kocyigit et al.
Journal of hazardous materials, 183(1-3), 334-340 (2010-08-10)
We presented the synthesis of a novel Schiff base and its complexation properties with some transition metal ions in this work. For this, the 2,3,4,6,7,10,11 hexahydroxytriphenylene (HHTP) as starting material was synthesized according to known procedure. Moreover, the 2-((2-chloroethylimino)methyl) phenol

Articles

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service