Merck
  • Mcl-1 is an important therapeutic target for oral squamous cell carcinomas.
Mcl-1 is an important therapeutic target for oral squamous cell carcinomas.
Oncotarget (2015-05-27)
Santanu Maji, Sabindra K Samal, Laxmipriya Pattanaik, Swagatika Panda, Bridget A Quinn, Swadesh K Das, Devanand Sarkar, Maurizio Pellecchia, Paul B Fisher, Rupesh Dash
ABSTRACT
Oral and oropharyngeal cancers are the sixth most common cancers worldwide. Despite intensive investigation, oral squamous cell carcinomas (OSCC) represent a clinical challenge resulting in significant morbidity and mortality. Resistance to cell death is common in OSCC and is often mediated by the Bcl-2 family proteins. Among all anti-apoptotic Bcl-2 family members, Mcl-1 functions as a major survival factor, particularly in solid cancers. Despite the confirmed importance of Mcl-1 in several neoplasms, the role of Mcl-1 in OSCC survival has yet to be explored. In this study, we found that knocking down Mcl-1 sensitized OSCC cells to ABT-737, which binds to Bcl-2/Bcl-xL but not Mcl-1. We report for the first time that a BH3 mimetic, Sabutoclax, which functions as an inhibitor of all anti-apoptotic Bcl-2 proteins, induced cancer-specific cell death in an Mcl-1-dependent manner through both apoptosis and toxic mitophagy. In vivo studies demonstrated that Sabutoclax alone decreased tumor growth in a carcinogen-induced tongue OSCC mouse model. In a combination regimen, Sabutoclax and COX-2 inhibitor, Celecoxib, synergistically inhibited the growth of OSCC in vitro and also significantly reduced OSCC tumor growth in vivo. Overall, these results identify Mcl-1 as a therapeutic prospective target in OSCC.
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human BNIP3
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Disodium succinate, ≥98%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mcl1
Sigma-Aldrich
MISSION® esiRNA, targeting human MCL1
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Retinoic acid p-hydroxyanilide, ≥95%
Sigma-Aldrich
4-Nitroquinoline N-oxide, ≥98%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bnip3