Skip to Content
Merck
All Photos(1)

Key Documents

T4069

Sigma-Aldrich

JC-1

≥97% purity, solid

Synonym(s):

5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethyl-imidacarbocyanine iodide, 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethylbenzimidazolocarbocyanine iodide, CBIC2(3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27Cl4IN4
CAS Number:
Molecular Weight:
652.23
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

JC-1, powder or solid (Crystals)

Assay

≥97%

form

powder or solid (Crystals)

technique(s)

titration: suitable

color

faint red to very dark red

ε (extinction coefficient)

≥175000 at 511-517 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[I-].CCN1C(=C\C=C\c2n(CC)c3cc(Cl)c(Cl)cc3[n+]2CC)/N(CC)c4cc(Cl)c(Cl)cc14

InChI

1S/C25H27Cl4N4.HI/c1-5-30-20-12-16(26)17(27)13-21(20)31(6-2)24(30)10-9-11-25-32(7-3)22-14-18(28)19(29)15-23(22)33(25)8-4;/h9-15H,5-8H2,1-4H3;1H/q+1;/p-1

InChI key

FYNNIUVBDKICAX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

A dual-emission potential-sensitive probe that can be used to measure mitochondrial membrane potential. JC-1 is a green-fluorescent (λex 520 nm) monomer at low membrane potential. At higher potentials, JC-1 forms red-fluorescent (λem 596 nm ) "J-aggregates," which exhibit broad excitation and very narrow emission spectra. The ratio of red to green fluorescence of JC-1 is dependent only on membrane potential, and not influenced by mitochondrial size, shape, or density.
The cyanine dye[1] JC-1 has been used to determine mitochondrial transmembrane potential.[2][3][4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junctophilin 3 expresses in pancreatic beta cells and is required for glucose-stimulated insulin secretion
Li L
Cell Death & Disease (2016)
Shumin Qin et al.
Frontiers in pharmacology, 10, 894-894 (2019-09-03)
Alcohol consumption affects gastric mucosa by multiple and complex mechanisms depending either by direct contact of ethanol or by indirect biological damage induced by its metabolite acetaldehyde. The present study aims at further investigating the mechanism of ethanol-induced gastric mucosa
Xianbin Cao et al.
Cellular signalling, 62, 109337-109337 (2019-06-08)
Radiation-induced tumor cells death is the theoretical basis of tumor radiotherapy. Death signaling disorder is the most important factor for radioresistance. However, the signaling pathway(s) leading to radiation-triggered cell death is (are) still not completely known. To better understand the
Chrysin induces cell apoptosis in human uveal melanoma cells via intrinsic apoptosis
Chunyan Xue
Oncology Letters (2016)
Sulfated polysaccharide-protein complex sensitizes doxorubicin-induced apoptosis of breast cancer cells in vitro and in vivo
Jie Wang
Experimental and Therapeutic Medicine (2016)

Articles

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

See All

Questions

  1. Mitochondria Staining Kit (CS0390)에 들어있는 1MG-JC-1 powder와 다른 제품인가요? 동일 농도로 DMSO이용해 녹였더니 Stock 색상이 다릅니다.

    1 answer
    1. Yes, the T4069, JC-1 product is also supplied in the Mitochndria Stain kit, CS0390. Both are prepared at 1 mg/mL in DMSO resulting in a red solution. Slight variations in color may be considered normal and the result of lot-to-lot variability. Marked differences in color should be discussed with Technical Service.

      Should this be the case, please navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquiries" under the Products Section and respond with all the required information so that a member of the Technical Service team can reach out to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service