Merck
  • Immunoaffinity chromatography purification and ultrahigh performance liquid chromatography tandem mass spectrometry determination of tetrodotoxin in marine organisms.
Immunoaffinity chromatography purification and ultrahigh performance liquid chromatography tandem mass spectrometry determination of tetrodotoxin in marine organisms.
Journal of agricultural and food chemistry (2015-03-11)
Xiaojun Zhang, Zhongyong Yan, Ying Wang, Tao Jiang, Jian Wang, Xiumei Sun, Yuanming Guo
ABSTRACT
A highly selective and sensitive method was developed for the determination of tetrodotoxin (TTX) in marine organisms by immunoaffinity chromatography (IAC) purification coupled with ultrahigh performance liquid chromatography tandem mass spectrometry (UPLC-MS/MS). An IAC column was prepared and used to cleanup the extracted samples. The operating conditions of the IAC column were optimized, and the capacity of new IAC column was found to be 1106 ng mL(-1), which was sufficient for TTX determination. The MS/MS conditions and UPLC mobile phase were also studied to optimize the operation conditions. Fortified marine organism samples at levels of 0.3-5.0 ng g(-1) were utilized, and the average recoveries were 86.5-103.6% with intra- and inter-day relative standard deviations less than 7.22 and 9.88%, respectively. The limits of detection and quantification were 0.1 and 0.3 ng g(-1), respectively. The method was later successfully applied for the determination of TTX in 100 marine organism samples collected from local markets.
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methyl Red solution, pH 4.2-6.2
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium sulfite-34S, 90 atom % 34S, 95% (CP)
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard