Skip to Content
Merck
All Photos(1)

Key Documents

437549

Sigma-Aldrich

Ethyl acetate

ACS reagent, ≥99.5%

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
grade:
ACS reagent
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)

grade

ACS reagent

Quality Level

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

product line

SAFC Hitech®

Assay

≥99.5%

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

dilution

(for analytical testing)

impurities

H2SO4, passes test (darkened)
≤0.0009 meq/g Titr. acid
≤0.2% water

evapn. residue

≤0.003%

color

APHA: ≤10

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

solubility

alcohol: soluble(lit.)
chloroform: soluble in salt form(lit.)

density

0.902 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl acetate, a carboxylate ester, is bio-friendly organic solvent with a wide range of industrial applications. Its synthesis by reactive distillation and by acceptorless dehydrogenative dimerization of ethanol has been explored. Its utility as a less toxic alternative to diethyl ether in the formalin-ether (F-E) sedimentation procedure for intestinal parasites has been investigated. Its ability as an acyl acceptor in the immobilized lipase-mediated preparation of biodiesel from crude vegetable oils has been examined. The complete degradation of ethyl acetate to CO2 using manganese octahedral molecular sieve (OMS-2) has been investigated.

Application

Ethyl acetate may be used in the following processes:
  • Preparation of thin films of TiO2 (titanium dioxide) on glass.
  • As an extraction medium in the multiresidue analysis of pesticide residues in fruit and vegetables.
  • Acetylation of primary amines to form amides in the presence of dimethyltin(IV) acetic acid distannoxane.

Legal Information

SAFC Hitech is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dynamics and control of an ethyl acetate reactive distillation column.
Vora N and Daoutidis P.
Industrial & Engineering Chemistry Research, 40(3), 833-849 (2001)
Investigation of ethyl acetate reactive distillation process.
Kenig EY, et al.
Chemical Engineering Science, 56(21), 6185-6193 (2001)
Catalytic Acetylation Amines with Ethyl Acetate.
Camacho-Camacho C, et al.
Main Group Metal Chemistry, 31(1-2), 13-20 (2008)
Manganese oxide OMS-2 as an effective catalyst for total oxidation of ethyl acetate.
Gandhe AR, et al.
Applied Catalysis. B, Environmental, 72(1), 129-135 (2007)
Anatase thin films on glass from the chemical vapor deposition of titanium (IV) chloride and ethyl acetate.
O'Neill SA, et al.
Chemistry of Materials, 15(1), 46-50 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service