Skip to Content
Merck
All Photos(1)

Key Documents

07-2190

Sigma-Aldrich

Dichloromethane

SAJ first grade, ≥99.0%

Synonym(s):

Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

SAJ first grade

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

1223 °F

expl. lim.

22 %

availability

available only in Japan

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aria Nouri et al.
Spine, 40(14), 1092-1100 (2015-04-22)
Ambispective study. To determine whether MRI parameters improve the predictive performance of a validated clinical prediction rule used to assess functional outcomes in surgical patients with DCM. Degenerative cervical myelopathy (DCM) is the most common cause of spinal cord dysfunction
Damian M Herz et al.
Brain : a journal of neurology, 138(Pt 6), 1658-1666 (2015-04-18)
Dopaminergic signalling in the striatum contributes to reinforcement of actions and motivational enhancement of motor vigour. Parkinson's disease leads to progressive dopaminergic denervation of the striatum, impairing the function of cortico-basal ganglia networks. While levodopa therapy alleviates basal ganglia dysfunction
Molamma P Prabhakaran et al.
Materials science & engineering. C, Materials for biological applications, 56, 66-73 (2015-08-08)
Advanced engineering of materials for the development of drug delivery devices provides scope for novel and versatile strategies for treatment of various diseases. 'Electrospraying' was used to prepare PLGA microparticles and further encapsulate the drug, metronidazole (Met) within the particles
Angharad M Roberts et al.
Science translational medicine, 7(270), 270ra6-270ra6 (2015-01-16)
The recent discovery of heterozygous human mutations that truncate full-length titin (TTN, an abundant structural, sensory, and signaling filament in muscle) as a common cause of end-stage dilated cardiomyopathy (DCM) promises new prospects for improving heart failure management. However, realization
K E Mauck et al.
Integrative and comparative biology, 54(2), 193-209 (2014-04-22)
Recent research suggests that plant viruses, and other pathogens, frequently alter host-plant phenotypes in ways that facilitate transmission by arthropod vectors. However, many viruses infect multiple hosts, raising questions about whether these pathogens are capable of inducing transmission-facilitating phenotypes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service