Merck
  • Intermediate filament protein accumulation in motor neurons derived from giant axonal neuropathy iPSCs rescued by restoration of gigaxonin.
Intermediate filament protein accumulation in motor neurons derived from giant axonal neuropathy iPSCs rescued by restoration of gigaxonin.
Human molecular genetics (2014-11-16)
Bethany L Johnson-Kerner, Faizzan S Ahmad, Alejandro Garcia Diaz, John Palmer Greene, Steven J Gray, Richard Jude Samulski, Wendy K Chung, Rudy Van Coster, Paul Maertens, Scott A Noggle, Christopher E Henderson, Hynek Wichterle
ABSTRACT
Giant axonal neuropathy (GAN) is a progressive neurodegenerative disease caused by autosomal recessive mutations in the GAN gene resulting in a loss of a ubiquitously expressed protein, gigaxonin. Gene replacement therapy is a promising strategy for treatment of the disease; however, the effectiveness and safety of gigaxonin reintroduction have not been tested in human GAN nerve cells. Here we report the derivation of induced pluripotent stem cells (iPSCs) from three GAN patients with different GAN mutations. Motor neurons differentiated from GAN iPSCs exhibit accumulation of neurofilament (NF-L) and peripherin (PRPH) protein and formation of PRPH aggregates, the key pathological phenotypes observed in patients. Introduction of gigaxonin either using a lentiviral vector or as a stable transgene resulted in normalization of NEFL and PRPH levels in GAN neurons and disappearance of PRPH aggregates. Importantly, overexpression of gigaxonin had no adverse effect on survival of GAN neurons, supporting the feasibility of gene replacement therapy. Our findings demonstrate that GAN iPSCs provide a novel model for studying human GAN neuropathologies and for the development and testing of new therapies in relevant cell types.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone LN-6, ascites fluid
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse