Skip to Content
Merck
All Photos(1)

Key Documents

SML0868

Sigma-Aldrich

Purmorphamine

≥98% (HPLC), powder, Hedgehog signaling activator

Synonym(s):

2-(1-Naphthoxy)-6-(4-morpholinoanilino)-9-cyclohexylpurine; 9-Cyclohexyl-N-[4-(4-morpholinyl)phenyl]-2-(1-naphthalenyloxy)-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H32N6O2
CAS Number:
Molecular Weight:
520.62
UNSPSC Code:
51111800
NACRES:
NA.77

Product Name

Purmorphamine, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

N7(CCOCC7)c1ccc(cc1)Nc2nc(nc5[n](cnc52)C6CCCCC6)Oc3c4c(ccc3)cccc4

InChI

1S/C31H32N6O2/c1-2-9-25(10-3-1)37-21-32-28-29(33-23-13-15-24(16-14-23)36-17-19-38-20-18-36)34-31(35-30(28)37)39-27-12-6-8-22-7-4-5-11-26(22)27/h4-8,11-16,21,25H,1-3,9-10,17-20H2,(H,33,34,35)

InChI key

FYBHCRQFSFYWPY-UHFFFAOYSA-N

Application

Purmorphamine has been used to study the consequences of upregulation of Hedgehog signalling pathway in Tupfel long fin strain Zebrafish.

Biochem/physiol Actions

Purmorphamine activates the Hedgehog (Hh) signaling pathway, directly binding to the Smoothened (Smo) receptor.
Purmorphamine activates the Hedgehog (Hh) signaling pathway, directly binding to the Smoothened (Smo) receptor. In various studies, purmorphamine has been shown to induce osteoblast differentiation, generate cardiomyocytes from human embryonic stem cells, increase the neuronal differentiation of a human striatal neural stem cell line, and to differentiate human umbilical cord mesenchymal stem cells (hUCMSCs) into motoneurons.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methylation microarray studies highlight PDGFA expression as a factor in biliary atresia.
Cofer Z C, et al.
PLoS ONE, 11(3), e0151521-e0151521 (2016)
Sara Brilha et al.
Scientific reports, 7(1), 16031-16031 (2017-11-24)
Central nervous system tuberculosis (CNS TB) has a high mortality and morbidity associated with severe inflammation. The blood-brain barrier (BBB) protects the brain from inflammation but the mechanisms causing BBB damage in CNS TB are uncharacterized. We demonstrate that Mycobacterium
Shabana Amanda Ali et al.
Arthritis & rheumatology (Hoboken, N.J.), 68(1), 127-137 (2015-09-01)
With no effective therapies to attenuate cartilage degeneration in osteoarthritis (OA), the result is pain and disability. Activation of hedgehog (HH) signaling causes changes related to the progression of OA, with higher levels of Gli-mediated transcriptional activation associated with increased
Roberto Simone et al.
EMBO molecular medicine, 10(1), 22-31 (2017-11-09)
Intronic GGGGCC repeat expansions in
Anna Drannik et al.
PloS one, 12(2), e0171668-e0171668 (2017-02-09)
Sonic hedgehog (Shh) is a morphogen essential to the developing nervous system that continues to play an important role in adult life by contributing to cell proliferation and differentiation, maintaining blood-brain barrier integrity, and being cytoprotective against oxidative and excitotoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service