Merck
  • Preparation, characterization and application of N-methylene phosphonic acid chitosan grafted magnesia-zirconia stationary phase.
Preparation, characterization and application of N-methylene phosphonic acid chitosan grafted magnesia-zirconia stationary phase.
Analytica chimica acta (2014-12-07)
Qing Wang, Jie Chen, Kun Huang, Xin Zhang, Li Xu, Zhi-guo Shi
ABSTRACT
A hydrophilic stationary phase (SP) was prepared through grafting N-methylene phosphonic acid chitosan on magnesia-zirconia particles (P-CTS-MgO-ZrO2) via Lewis acid-base interaction. The resulting material was characterized by thermogravimetric analysis, Fourier transform infrared spectroscopy, scanning electron microscope and nitrogen adsorption analysis. The chromatographic performance of P-CTS-MgO-ZrO2 was systemically evaluated by studying effect of acetonitrile content, pH and buffer concentration in the mobile phase. The results demonstrated that the novel SP provided hydrophilic, electrostatic-repulsion and ion-exchange interactions. Compared to the bare MgO-ZrO2, P-CTS-MgO-ZrO2 exhibited superior peak shape, reasonable resolution and reduced analysis time in separation of basic analytes. Besides, remarkable resolving power of acids, i.e. six non-steroidal anti-inflammatory drugs which failed to be eluted from the bare MgO-ZrO2, was obtained with the theoretical plate number (N/m) of 4653-31313, asymmetry factor <1.21 and the resolution of 1.6-3.4. Finally, P-CTS-MgO-ZrO2 SP was applied to separate monosaccharides, phospholipids and peptides. P-CTS-MgO-ZrO2 was a promising hydrophilic SP for wide applications.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
D-(−)-Ribose, 98%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Ribose, suitable for cell culture, BioReagent
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Galactose, European Pharmacopoeia (EP) Reference Standard
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Xylose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%