Skip to Content
Merck
All Photos(1)

Key Documents

63423

Sigma-Aldrich

Maltose solution

BioReagent, for molecular biology, ~20% in H2O

Synonym(s):

4-O-α-D-Glucopyranosyl-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93798
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

grade

Molecular Biology
for molecular biology

product line

BioReagent

form

liquid (clear)

concentration

~20% in H2O

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

color

colorless

refractive index

n20/D 1.361

pH

4.0-6.5 (25 °C)

anion traces

chloride (Cl-): ≤25 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.05 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤20 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤10 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.08
λ: 280 nm Amax: 0.07

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5-,6+,7-,8-,9-,10-,11?,12-/m1/s1

InChI key

GUBGYTABKSRVRQ-PICCSMPSSA-N

Application

D-Maltose, an α(1,4) linked disaccharide of D-glucose, is used as a substrate for the identification, differentiation and characterization of enzymes such as maltase(s); maltose α-D-glucosyltransferase(s); maltose-transporting ATPase(s); maltose O-acetyltransferase(s) and maltose epimerase(s) and phosphorylase(s). D-Maltose is used to study maltose-binding proteins and disaccharide transport systems.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thaddeus C Deiss et al.
Journal of immunology (Baltimore, Md. : 1950), 203(5), 1265-1275 (2019-07-26)
The loci encoding B and T cell Ag receptors are generally distinct in commonly studied mammals, with each receptor's gene segments limited to intralocus, cis chromosomal rearrangements. The nurse shark (Ginglymostoma cirratum) represents the oldest vertebrate class, the cartilaginous fish
Aylin W Sahin et al.
Food & function, 10(8), 4985-4997 (2019-07-28)
Sugar reduction in sweet baked goods is one of the most popular trends on the food market. However, reducing sugar without lowering the product quality with respect to sweetness, texture and microbial shelf life is challenging. Sugar alcohols are one
Adarsh K Gupta et al.
Virus research, 269, 197640-197640 (2019-06-21)
Triticum mosaic virus (TriMV) is the exemplar strain of the type species of the genus Poacevirus in the family Potyviridae infecting wheat in the Great Plains region of the USA. Previously, we reported that the P1 protein of TriMV is
Nadja Jessberger et al.
Food microbiology, 84, 103276-103276 (2019-08-20)
The diarrheal type of food poisoning caused by enteropathogenic Bacillus cereus has been linked to various exotoxins. Best described are the non-hemolytic enterotoxin (Nhe), hemolysin BL (Hbl), and cytotoxin K (CytK). Due to the ubiquitous prevalence of B. cereus in
Marie Horká et al.
ACS infectious diseases, 5(8), 1348-1356 (2019-06-28)
Cellulose-based preparative isoelectric focusing was used for preseparation and concentration of uropathogens Staphylococcus aureus, Escherichia coli, Enterococcus faecalis, Staphylococcus epidermidis, Candida albicans, and Candida parapsilosis in a urine sample containing a high concentration of human serum albumin. For the visibility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service