Skip to Content
Merck
All Photos(1)

Key Documents

47673

Sigma-Aldrich

Formaldehyde solution

tested according to Ph. Eur.

Synonym(s):

Formaldehydi solutio 35 per centum, Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

form

liquid

autoignition temp.

572 °F

contains

9.0-15.0 % (v/v) methanol as stabilizer

concentration

25-50%

solubility

water: soluble

density

1.09 g/mL at 25 °C (lit.)

suitability

suitable for manufacturing use (Laboratory chemicals)

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formaldehyde solution has been used to fix labeled cells during flow cytometry.[1]

Biochem/physiol Actions

Formaldehyde is the simplest aldehyde that denatures the bihelical regions of RNA and converts the polynucleotides into random coils.[2] It is a genotoxic substance that significantly induces DNA-protein crosslinks (DPC), sister-chromatid exchanges, micronuclei formation and leads to cytotoxicity.[3] It also induces tumors in the nasal epithelium of rats and supposed to be a human carcinogen.

Caution

may become turbid on storage

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

133.0 °F - closed cup

Flash Point(C)

56.11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dependence of the sedimentation coefficient on molecular weight of RNA after reaction with formaldehyde.
H Boedtker
Journal of molecular biology, 35(1), 61-70 (1968-07-14)
J L Wilson et al.
Journal of immunology (Baltimore, Md. : 1950), 163(12), 6365-6370 (1999-12-10)
NK cells have the capacity to spontaneously kill tumor cell lines, in particular cell lines of hemopoietic origin. In contrast, they do not generally kill nontransformed autologous cells. However, here we demonstrate that short-term activated polyclonal human NK cells, as
O Merk et al.
Environmental and molecular mutagenesis, 32(3), 260-268 (1998-11-14)
Formaldehyde (FA) is a genotoxic substance, induces tumors in the nasal epithelium of rats, and is suspected to be a human carcinogen. As a primary DNA lesion, FA induces DNA-protein crosslinks (DPC) and the formation of DPC has been used
Cindy S Ma et al.
The Journal of experimental medicine, 205(7), 1551-1557 (2008-07-02)
Hyper-immunoglobulin E syndrome (HIES) is a primary immune deficiency characterized by abnormal and devastating susceptibility to a narrow spectrum of infections, most commonly Staphylococcus aureus and Candida albicans. Recent investigations have identified mutations in STAT3 in the majority of HIES
Stephanie Belloc et al.
Fertility and sterility, 101(6), 1588-1593 (2014-04-03)
To evaluate sperm DNA fragmentation in normozoospermic male partners of couples undergoing infertility evaluation. Retrospective cohort study. Clinical andrology laboratory. A total of 1,974 consecutive normozoospermic men selected from a larger cohort of 4,345 consecutive, nonazoospermic men presenting for infertility

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service