Merck
  • Amplified cold transduction in native nociceptors by M-channel inhibition.
Amplified cold transduction in native nociceptors by M-channel inhibition.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-10-18)
Irina Vetter, Alexander Hein, Simon Sattler, Sabine Hessler, Filip Touska, Elisangela Bressan, Andres Parra, Ulrich Hager, Andreas Leffler, Stepana Boukalova, Matthias Nissen, Richard J Lewis, Carlos Belmonte, Christian Alzheimer, Tobias Huth, Viktorie Vlachova, Peter W Reeh, Katharina Zimmermann
ABSTRACT
Topically applied camphor elicits a sensation of cool, but nothing is known about how it affects cold temperature sensing. We found that camphor sensitizes a subpopulation of menthol-sensitive native cutaneous nociceptors in the mouse to cold, but desensitizes and partially blocks heterologously expressed TRPM8 (transient receptor potential cation channel subfamily M member 8). In contrast, camphor reduces potassium outward currents in cultured sensory neurons and, in cold nociceptors, the cold-sensitizing effects of camphor and menthol are additive. Using a membrane potential dye-based screening assay and heterologously expressed potassium channels, we found that the effects of camphor are mediated by inhibition of Kv7.2/3 channels subtypes that generate the M-current in neurons. In line with this finding, the specific M-current blocker XE991 reproduced the cold-sensitizing effect of camphor in nociceptors. However, the M-channel blocking effects of XE991 and camphor are not sufficient to initiate cold transduction but require a cold-activated inward current generated by TRPM8. The cold-sensitizing effects of XE991 and camphor are largest in high-threshold cold nociceptors. Low-threshold corneal cold thermoreceptors that express high levels of TRPM8 and lack potassium channels are not affected by camphor. We also found that menthol--like camphor--potently inhibits Kv7.2/3 channels. The apparent functional synergism arising from TRPM8 activation and M-current block can improve the effectiveness of topical coolants and cooling lotions, and may also enhance TRPM8-mediated analgesia.
Sigma-Aldrich
(±)-Camphor, SAJ first grade, ≥96.0%
Sigma-Aldrich
(1R)-(+)-Camphor, 98%
Sigma-Aldrich
DL-Menthol, ≥95%, FCC, FG
Sigma-Aldrich
(±)-Camphor, ≥95.5%
Sigma-Aldrich
Camphor, 96%
Supelco
(−)-Camphor, analytical standard
Sigma-Aldrich
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Sigma-Aldrich
(±)-Menthol, racemic, ≥98.0% (GC)
Sigma-Aldrich
(±)-Camphor, purum, synthetic, ≥95.0% (GC)
Camphor (dl), primary reference standard
Menthol, European Pharmacopoeia (EP) Reference Standard
Camphor (racemic), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Menthol, 99%
Sigma-Aldrich
(1S)-(−)-Camphor, 95%
Sigma-Aldrich
D-Camphor, ≥97%, FG
Supelco
D-Camphor, Pharmaceutical Secondary Standard; Certified Reference Material