Skip to Content
Merck
All Photos(3)

Key Documents

857300

Sigma-Aldrich

(1R)-(+)-Camphor

98%

Synonym(s):

(+)-Camphor, (1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2042745
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.24 (vs air)

Quality Level

vapor pressure

4 mmHg ( 70 °C)

Assay

98%

form

powder

optical activity

[α]25/D +44.1°, c = 10 in ethanol

autoignition temp.

870 °F

expl. lim.

3.5 %

mp

179-181 °C (lit.)

functional group

ketone

SMILES string

C[C@@]1(CC2)C(C)(C)[C@H]2CC1=O

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Application

(1R)-(+)-Camphor has been used as a standard during the enantioselective gas chromatography-mass spectrometry (GC-MS) analysis of the constituents of Achillea ligustica essential oil.
It may also be used to synthesize:
  • {N,N′-bis[(1R,2R,4R)-1,7,7-trimethylbicyclo[2.2.1]hept-2-yl]-1,2-ethanediamine}, a C2-symmetrical diamine
  • chiral oxazaborolidines
  • (-)-(1R,2R)-1,2-dihydroxy-3,3-dimethylnorbornane
Chiral intermediate and auxiliary.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Target Organs

Lungs

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Asymmetric reduction of prochiral ketones using in situ generated oxazaborolidines derived from amino alcohols of (1R)-camphor as catalysts.
Santhi V and Rao JM
Tetrahedron Asymmetry, 11(17), 3553-3560 (2000)
Synthesis of homochiral 1, 2-diols from (-)-fenchone and (+)-camphor.
Martinez AG, et al.
Tetrahedron Asymmetry, 5(7), 1373-1376 (1994)
Synthesis of C 2-symmetrical diamine based on (1R)-(+)-camphor and application to oxidative aryl coupling of naphthols.
Caselli A, et al.
Tetrahedron Asymmetry, 14(11), 1451-1454 (2003)
Synthetic Communications, 23, 437-437 (1993)
The Journal of Organic Chemistry, 56, 1185-1185 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service