Skip to Content
Merck
All Photos(1)

Key Documents

C0405000

Camphor (racemic)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
1907611
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

5.2 (vs air)

vapor pressure

4 mmHg ( 70 °C)

API family

camphor

expl. lim.

3.5 %

manufacturer/tradename

EDQM

bp

204 °C (lit.)

mp

175-177 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@](CC1=O)(CC2)C(C)(C)[C@]12C

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Camphor (racemic) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.9 °F - closed cup

Flash Point(C)

64.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyuki Yamashita et al.
Pharmaceutical research, 30(1), 70-80 (2012-08-22)
Although a number of studies have reported that cocrystals can form by heating a physical mixture of two components, details surrounding heat-induced cocrystal formation remain unclear. Here, we attempted to clarify the thermal behavior of a physical mixture and cocrystal
I Jiménez-Díaz et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 936, 80-87 (2013-09-06)
UV-filters are widely used in many personal care products and cosmetics. Recent studies indicate that some organic UV-filters can accumulate in biota and act as endocrine disruptors, but there are few studies on the occurrence and fate of these compounds
Yali V Zhang et al.
Nature neuroscience, 16(10), 1468-1476 (2013-09-10)
Animals tend to reject bitter foods. However, long-term exposure to some unpalatable tastants increases acceptance of these foods. Here we show that dietary exposure to an unappealing but safe additive, camphor, caused the fruit fly Drosophila melanogaster to decrease camphor
Skrollan Stockinger et al.
Journal of chromatography. A, 1269, 346-351 (2012-08-23)
Novel 3-(perfluoroalkanoyl)-(1R)-camphorate nickel complexes immobilized to poly(dimethylsiloxane) phases are presented. Immobilized 3-(perfluoroalkanoyl)-(1R)-camphorate nickel complexes with a trifluoromethyl (CF(3); nickel(II)-bis[(1R,4S)-3-trifluoromethanoyl-10-propylenoxycamphor]-polysiloxane Ni(tfpc)(2)@PS) and a heptafluoropropyl-substituent (C(3)F(7); nickel(II)-bis[(1R,4S)-3-heptafluorobutanoyl-10-propylenoxycamphor]-polysiloxane Ni(hfpc)(2)@PS) were synthesized, characterized and immobilized to polysiloxane. Ni(hfpc)(2)@PS was immobilized with a selector content
M Shehata et al.
Archives of toxicology, 86(9), 1423-1429 (2012-05-23)
Phenol has been traditionally used in dental treatment as a sedative for the pulp or as disinfectant for carious cavity and root canal. However, phenol is regarded as a mutagenic and carcinogenic agent and its use in dental practice is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service