Skip to Content
Merck
All Photos(1)

Key Documents

63670

Sigma-Aldrich

(±)-Menthol

racemic, ≥98.0% (GC)

Synonym(s):

Menthol, Racemic menthol, 2-Isopropyl-5-methylcyclohexanol, Hexahydrothymol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
3194263
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.8 mmHg ( 20 °C)

Assay

≥98.0% (GC)

form

solid

quality

racemic

bp

216 °C (lit.)

mp

34-36 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)C1CCC(C)CC1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3

InChI key

NOOLISFMXDJSKH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

(±)-Menthol (racemic menthol) can be synthesized via hydrogenation of citral.

Application

(±)-Menthol (racemic menthol) can be transformed to (-)-menthyl acetate by using Candida rugosa lipase(CRL) and lipase PS(from Pseudomonas cepacia) as catalysts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

204.8 °F

Flash Point(C)

96 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Design, scale up and safe piloting of thymol hydrogenation and menthol racemisation"
Dudas J and Hanika J
Chemical Engineering Research and Design, 87(01), 83-90 (2009)
"Lipase-catalyzed resolution of p-menthan-3-ols monoterpenes: preparation of the enantiomer-enriched forms of menthol, isopulegol, trans-and cis-piperitol, and cis-isopiperitenol"
Serra S, et al.
Tetrahedron Asymmetry, 14(21), 3313-3319 (2003)
Joanne D'Silva et al.
American journal of preventive medicine, 43(5 Suppl 3), S242-S248 (2012-10-25)
Menthol cigarettes account for 25% of the market in the U.S. The Food and Drug Administration currently is considering regulatory action on tobacco products, including a ban on menthol cigarettes. With 39% of menthol smokers reporting that they would quit
Brandon J Henderson et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(10), 2957-2974 (2016-03-11)
Upregulation of β2 subunit-containing (β2*) nicotinic acetylcholine receptors (nAChRs) is implicated in several aspects of nicotine addiction, and menthol cigarette smokers tend to upregulate β2* nAChRs more than nonmenthol cigarette smokers. We investigated the effect of long-term menthol alone on
María Ll Valero et al.
PloS one, 7(12), e51825-e51825 (2012-12-20)
Overexpression of the cation-permeable channel TRPM8 in prostate cancers might represent a novel opportunity for their treatment. Inhibitors of TRPM8 reduce the growth of prostate cancer cells. We have used two recently described and highly specific blockers, AMTB and JNJ41876666

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service