Merck
  • Adult granulosa cell tumours (GCT): clinicopathological outcomes including FOXL2 mutational status and expression.
Adult granulosa cell tumours (GCT): clinicopathological outcomes including FOXL2 mutational status and expression.
Gynecologic oncology (2013-09-07)
Roseanne Rosario, Michelle Wilson, Wei-Tzu Cheng, Kathryn Payne, Paul A Cohen, Peter Fong, Andrew N Shelling
ABSTRACT
The aim of this research was to use nucleic acids isolated from formalin-fixed paraffin-embedded (FFPE) tissue to investigate the diagnostic potential and prognostic significance of FOXL2 in adult-type GCTs, particularly as a marker of identifying early stage patients that are likely to relapse. We performed a retrospective review of GCT patients referred to the Auckland Gynae-Oncology Multidisciplinary Team from 1955 to 2012. Baseline characteristics, clinical course, histopathology and survival data was recorded. Using nucleic acids extracted from FFPE tumour blocks, FOXL2 mutation status and expression was determined by DNA sequencing and RT-qPCR, respectively, and correlated with clinical data. 57 adult GCT patients were identified, however FFPE tumour blocks were available for only 37 of these patients. Sequencing results confirmed the presence of the FOXL2 mutation in 70% of patients. FOXL2 mutation positive adult tumours showed a trend towards higher FOXL2 expression than wildtype adult tumours, particularly in stage I patients (p=0.051). In addition, patients with homozygous FOXL2 mutations had a significantly higher relapse rate (p=0.04). There was no significant correlation between FOXL2 mutation status or FOXL2 expression and any other clinical variables. FFPE tumour blocks are a valuable resource of molecular information, especially when studying rare tumours such as GCTs. The FOXL2 mutation appears to have some diagnostic potential, however additional work in a larger cohort needs to be completed to confirm the prognostic significance of this gene mutation, and its expression.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, 10%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.