Skip to Content
Merck
All Photos(1)

Key Documents

11-0705

Sigma-Aldrich

Formaldehyde neutral buffer solution

3.7% formaldehyde in H2O

Synonym(s):

Formaldehyde solution, Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

form

liquid

autoignition temp.

572 °F

availability

available only in Japan

concentration

3.7% formaldehyde in H2O

pH

7.10-7.30 (20 °C)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 1B - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gahana Advani et al.
Cell communication and signaling : CCS, 15(1), 29-29 (2017-08-09)
C-terminal Src kinase (Csk) and Csk-homologous kinase (Chk) are the major endogenous inhibitors of Src-family kinases (SFKs). They employ two mechanisms to inhibit SFKs. First, they phosphorylate the C-terminal tail tyrosine which stabilizes SFKs in a closed inactive conformation by
D H Castrillon et al.
The American journal of surgical pathology, 25(10), 1225-1230 (2001-11-02)
The p57KIP2 protein is a cell cycle inhibitor and tumor suppressor encoded by a strongly paternally imprinted gene. We explored the utility of p57KIP2 as a diagnostic marker in hydatidiform mole, a disease likely the result of abnormal dosage and
Jing Lu et al.
Biochimica et biophysica acta, 1830(8), 4102-4116 (2013-05-01)
Chronic formaldehyde exposure leads to memory impairment and abnormal elevation of endogenous formaldehyde has been found in the brains of Alzheimer's disease (AD) patients. Hyperphosphorylated Tau protein with subsequent aggregates as neurofibrillary tangles (NFTs) is one of the typical pathological
G R Moore et al.
Neurology, 55(10), 1506-1510 (2000-11-30)
To determine the pathologic basis of areas not exhibiting signal of the short-T2 component of the T2 relaxation distribution in MS, as studied in formalin-fixed brain. A myelin-specific MRI signal would be of great importance in assessing demyelination in patients
Roseanne Rosario et al.
Gynecologic oncology, 131(2), 325-329 (2013-09-07)
The aim of this research was to use nucleic acids isolated from formalin-fixed paraffin-embedded (FFPE) tissue to investigate the diagnostic potential and prognostic significance of FOXL2 in adult-type GCTs, particularly as a marker of identifying early stage patients that are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service