Merck
  • Free amino acid and dipeptide changes in the body fluids from Alzheimer's disease subjects.
Free amino acid and dipeptide changes in the body fluids from Alzheimer's disease subjects.
Amino acids (2006-10-13)
A N Fonteh, R J Harrington, A Tsai, P Liao, M G Harrington
ABSTRACT
Our aim was to determine changes in free amino acid (FAA) and dipeptide (DP) concentrations in probable Alzheimer's disease (pAD) subjects compared with control (CT) subjects using liquid chromatography and electrospray ionization tandem mass spectrometry (LCMS2). We recruited gender- and age-matched study participants based on neurological and neuropsychological assessments. We measured FAAs and DPs in cerebrospinal fluid (CSF), plasma and urine using LCMS2 with selected reaction monitoring (SRM). Imidazole-containing FAAs (histidine, methyl-histidine), catecholamines (L-DOPA and dopamine), citrulline, ornithine, glycine and antioxidant DPs (carnosine and anserine) accounted for the major changes between CT and pAD. Carnosine levels were significantly lower in pAD (328.4 +/- 91.31 nmol/dl) than in CT plasma (654.23 +/- 100.61 nmol/dl). In contrast, L-DOPA levels were higher in pAD (1400.84 +/- 253.68) than CT (513.10 +/- 121.61 nmol/dl) plasma. These data underscore the importance of FAA and DP metabolism in the pathogenesis of AD. Since our data show changes in antioxidants, neurotransmitters and their precursors, or FAA associated with urea metabolism in pAD compared with CT, we propose that manipulation of these metabolic pathways may be important in preventing AD progression.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Mercury, JIS special grade, ≥99.5%
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Mercury, SAJ first grade, ≥99.5%
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
SAFC
Glycine
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
myo-Inositol, ≥99%
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)