Skip to Content
Merck
All Photos(1)

Key Documents

19-1510

Sigma-Aldrich

Mercury

SAJ first grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Hg
CAS Number:
Molecular Weight:
200.59
MDL number:
UNSPSC Code:
12141726
PubChem Substance ID:
grade:
SAJ first grade
form:
liquid

grade

SAJ first grade

vapor density

7 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 126 °C)

Assay

≥99.5%

form

liquid

reaction suitability

reagent type: catalyst
core: mercury

availability

available only in Japan

resistivity

95.8 μΩ-cm, 20°C

bp

356.6 °C (lit.)

mp

−38.87 °C (lit.)

storage temp.

15-25°C

SMILES string

[Hg]

InChI

1S/Hg

InChI key

QSHDDOUJBYECFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Okechukwu S Ogah et al.
JACC. Heart failure, 2(3), 250-259 (2014-06-24)
The aim of this study was to determine the contemporary profile, clinical characteristics, and intrahospital outcomes of acute heart failure (AHF) in an African urban community. There are limited data on the current burden and characteristics of AHF in Nigerian
Alexandre J Poulain et al.
The ISME journal, 9(12), 2541-2550 (2015-06-10)
Mercury (Hg) is an anthropogenic pollutant that is toxic to wildlife and humans, but the response of remote ecosystems to globally distributed Hg is elusive. Here, we use DNA extracted from a dated sediment core to infer the response of
James P K Rooney
Toxicology, 234(3), 145-156 (2007-04-06)
Mercury has been a known as a toxic substance for centuries. Whilst the clinical features of acute mercury poisoning have been well described, chronic low dose exposure to mercury remains poorly characterised and its potential role in various chronic disease
Ekim Seven et al.
Journal of hypertension, 32(7), 1488-1494 (2014-05-09)
The adipocytokines, leptin, adiponectin, and interleukin-6, which stimulate liver C-reactive protein (CRP) production, are regarded as potential candidate intermediates between adipose tissue and overweight-induced hypertension. We examined the associations between leptin, adiponectin, and CRP levels with both prevalent and 5-year
Saul Miller et al.
American journal of kidney diseases : the official journal of the National Kidney Foundation, 62(1), 135-138 (2013-04-23)
Kidney injury from mercury is known to cause dose-related tubular dysfunction and idiosyncratic nephrotic syndrome according to various case reports. Motivated by a patient with subacute-onset nephrotic syndrome, histologic features of secondary focal segmental glomerulosclerosis, and concurrent mercury toxicity, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service