Merck
  • Immunocytochemistry performed on the cell-transferred direct smears of the fine-needle aspirates: a comparison study with the corresponding formalin-fixed paraffin-embedded tissue.
Immunocytochemistry performed on the cell-transferred direct smears of the fine-needle aspirates: a comparison study with the corresponding formalin-fixed paraffin-embedded tissue.
American journal of clinical pathology (2013-05-22)
Howard H Wu, Kelly J Jones, Harvey M Cramer
ABSTRACT
Immunocytochemistry (ICC) performed on the cell-transferred cytologic smears (CTCS) of fine-needle aspiration (FNA) is useful when the cell blocks lack adequate material. The comparison of the ICC results from the CTCS of FNA with the corresponding formalin-fixed paraffin-embedded tissue (FFPE) has not been reported previously. We applied 12 commonly used ICC antibodies on 160 pieces of ethanol-fixed, cell-transferred Papanicolaou-stained smears obtained from 42 FNA specimens and compared the staining results with the corresponding FFPE on which the same panel of immunostains was performed. Of the 160 pieces of transferred materials, only 3 (1.9%) were lost during specimen processing. In total, 153 of 157 (97.5%) showed staining results that agreed with the corresponding FFPE, including 78 of 81 positive staining and 75 of 76 negative staining cases. ICC performed on the cell-transferred FNA smears is reliable and shows staining results highly comparable with the corresponding FFPE tissue.
Product Description
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)