Merck
  • Semi-automated liquid chromatography-mass spectrometric imaging platform for enhanced detection and improved data analysis of complex peptides.
Semi-automated liquid chromatography-mass spectrometric imaging platform for enhanced detection and improved data analysis of complex peptides.
Journal of chromatography. A (2013-04-30)
Zichuan Zhang, Shan Jiang, Lingjun Li
ABSTRACT
A semi-automated analytical platform featuring the coupling of monolithic reversed-phase liquid chromatography (RPLC) to matrix-assisted laser desorption/ionization mass spectrometric imaging (MALDI MSI) has been developed and evaluated. This is the first time that LC separation is readily coupled to MS imaging detection for the analysis of complex peptide mixtures both qualitatively and quantitatively. Methacrylate-based monolithic column with C12 functional groups is fabricated for fast RPLC separation. The LC flow and matrix flow are collected on a commercially available MALDI plate which is mechanically controlled and analyzed with MALDI MSI subsequently. Both tryptic peptides digested from bovine serum albumin (BSA) and endogenous neuropeptides extracted from the blue crab Callinectes sapidus are analyzed with this novel LC-MSI platform. Compared with regular offline LC fractionation coupled with MALDI MS detection, LC-MSI exhibits significantly increased MS signal intensity due to retaining of temporal resolution from separation dimension via continuous sampling, which results in increased number of peptides detected and accurate quantitation. In addition, imaging signals enable improved data analysis based on either mass-to-charge ratio or retention time, which is extremely beneficial for the analysis of complex analytes. These findings have demonstrated the potential of employing LC-MSI platform for enhanced proteomics and peptidomics studies.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)