Merck
  • Shear bond strength to enamel and flexural strength of different fiber-reinforced composites.
Shear bond strength to enamel and flexural strength of different fiber-reinforced composites.
The journal of adhesive dentistry (2013-03-28)
Jelena Juloski, Milos Beloica, Cecilia Goracci, Nicoletta Chieffi, Agostino Giovannetti, Alessandro Vichi, Zoran R Vulicevic, Marco Ferrari
ABSTRACT
To assess the shear bond strength to unground human enamel (ESBS) and flexural strength (FS) of different reinforcing fibers used in combination with a flowable composite resin. For ESBS testing, 90 human molars were selected and randomly divided into 9 groups (n = 10) according to the reinforcing fiber to be tested: 1. RTD Quartz Splint additionally impregnated at chairside with Quartz Splint Resin (RTD); 2. RTD Quartz Splint without additional impregnation; 3. Ribbond-THM (Ribbond) impregnated with OptiBond FL Adhesive; 4: Ribbond Triaxial (Ribbond) impregnated with OptiBond FL Adhesive; 5. Connect (Kerr) impregnated with OptiBond FL Adhesive; 6. Construct (Kerr) impregnated with Opti- Bond FL Adhesive; 7. everStick PERIO (Stick Tech); 8. everStick C&B (Stick Tech); 9. nonreinforced composite Premise flowable (Kerr). Cylinders of flowable composite reinforced with the fibers were bonded to the intact buccal surface of the teeth. After 24 h of storage, shear loading was performed until failure occurred. FS was assessed performing three-point bending test according to ISO Standard 4049/2000. ESBS and FS data were analyzed using one-way ANOVA, followed by Tukey's HSD test for post-hoc comparisons (p < 0.05). For each group, the ESBS and FS, respectively, in MPa were: 1. 17.07 ± 4.52 and 472.69 ± 30.49; 2. 14.98 ± 3.92 and 441.77 ± 61.43; 3. 18.59 ± 5.67 and 186.89 ± 43.89; 4. 16.74 ± 6.27 and 314.41 ± 148.52; 5. 14.38 ± 4.14 and 223.80 ± 77.35; 6. 16.00 ± 5.55 and 287.62 ± 85.91; 7. 16.42 ± 3.67 and 285.35 ± 39.68; 8. 23.24 ± 5.81 and 370.46 ± 29.26; 9. 12.58 ± 4.76 and 87.75 ± 22.87. For most fibers, no significant difference in ESBS was found compared to the control group, except for everStick C&B, which yielded higher ESBS. Nonreinforced composite exhibited the lowest FS, while all fibers positively affected the FS. Fiber reinforcement of flowable composite does not affect its ESBS. The flexural strength of FRCs is significantly influenced by fiber composition and pattern.
Sigma-Aldrich
Silicon dioxide, ~99%, 0.5-10 μm (approx. 80% between 1-5 μm)
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Sand, white quartz, CP, beads
Polyethylene (LDPE), ERM®, certified reference material
Sigma-Aldrich
Sand, 50-70 mesh particle size