Skip to Content
Merck
All Photos(3)

Key Documents

429015

Sigma-Aldrich

Polyethylene

Ultra-high molecular weight, average Mw 3,000,000-6,000,000

Synonym(s):

PE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

coefficient of friction: 0.20 - 0.25

(Cr-plated steel, 23°C, static, ASTM D 1894)

form

powder

mol wt

average Mw 3,000,000-6,000,000

hardness

52 (Rockwell R, ASTM D 785)

transition temp

softening point 136 °C (Vicat, ASTM D 1525B)
Tm 138 °C (DSC)

density

0.94 g/mL at 25 °C

SMILES string

C=C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bearings, gears, bushings and other moving parts.

Features and Benefits

Self-lubricating extraordinary wear properties.
Abrasion, corrosion and impact resistance exceeds many conventional materials, including steel and expensive metals and alloys.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eliandra Mirlei Rossi et al.
Journal of infection in developing countries, 7(3), 229-234 (2013-03-16)
Contaminated sponges might lead to cross-contamination in kitchens since they can transfer microorganisms to surfaces where microorganisms can survive for hours or days and contaminate food. The main objective of this study was to evaluate the transfer and the survival
Morteza Meftah et al.
The Journal of bone and joint surgery. American volume, 95(13), 1193-1197 (2013-07-05)
Ceramic femoral heads produce less wear of the opposing polyethylene than do metal femoral heads in wear simulation studies. This is a matched-pair analysis of the wear of ceramic and metal femoral heads on conventional polyethylene in uncemented total hip
Jelena Juloski et al.
The journal of adhesive dentistry, 15(2), 123-130 (2013-03-28)
To assess the shear bond strength to unground human enamel (ESBS) and flexural strength (FS) of different reinforcing fibers used in combination with a flowable composite resin. For ESBS testing, 90 human molars were selected and randomly divided into 9
Lisa K McCarthy et al.
Pediatrics, 132(1), e135-e141 (2013-06-19)
Hypothermia on admission to the NICU is associated with increased mortality in preterm infants. Many newborns are hypothermic on admission despite using polyethylene bags (PBs). Using exothermic mattresses (EMs) in addition to PBs may reduce hypothermia but increase hyperthermia. We
Mahtab Memarpour et al.
The journal of adhesive dentistry, 15(1), 85-91 (2013-03-28)
To evaluate the clinical performance of polyethylene fiber for anterior primary teeth restorations in children after 2.5 years, according to modified Ryge criteria. Fifty-five maxillary incisors with extensive caries in 24 Iranian patients (mean age 4.2 years) were restored. All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service