Skip to Content
Merck
All Photos(4)

Key Documents

S5631

Sigma-Aldrich

Silicon dioxide

~99%, 0.5-10 μm (approx. 80% between 1-5 μm)

Synonym(s):

Sand, white quartz, Quartz, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
SB.52
Pricing and availability is not currently available.

Assay

~99%

Quality Level

form

powder

particle size

0.5-10 μm (approx. 80% between 1-5 μm)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Historically called amorphous.

Preparation Note

A naturally occurring microcrystalline silica which has been finely ground.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alida Kent et al.
Journal of tropical medicine, 2013, 324140-324140 (2013-11-07)
Cutaneous leishmaniasis (CL) is an emerging disease in Suriname, with at least 200 cases per year. Little is known about the biology of CL in the country. The most important parasite species is Leishmania Viannia guyanensis, but possible vectors and
Yan Liu et al.
Respiratory research, 18(1), 38-38 (2017-02-23)
Myofibroblasts play a major role in the synthesis of extracellular matrix (ECM) and the stimulation of these cells is thought to play an important role in the development of silicosis. The present study was undertaken to investigate the anti-fibrotic effects
Jian-Feng Li et al.
Plant methods, 6(1), 1-1 (2010-02-26)
Research in plant molecular biology involves DNA purification on a daily basis. Although different commercial kits enable convenient extraction of high-quality DNA from E. coli cells, PCR and agarose gel samples as well as plant tissues, each kit is designed
Haijun Liu et al.
American journal of physiology. Lung cellular and molecular physiology, 310(2), L121-L132 (2015-11-27)
Silicosis is a systemic disease caused by inhaling silicon dioxide (SiO2). Phagocytosis of SiO2 in the lungs initiates an inflammatory cascade that results in fibroblast proliferation and migration followed by fibrosis. According to previous data from our laboratory, monocyte chemotactic
Outi Leppäranta et al.
American journal of respiratory cell and molecular biology, 48(4), 448-455 (2012-12-22)
Idiopathic pulmonary fibrosis (IPF) is a progressive lung disease with a poor prognosis and very few therapeutic options. On the molecular level, patients with IPF have increased amounts of the bone morphogenetic protein (BMP) inhibitor gremlin in their lungs, which

Questions

1–3 of 3 Questions  
  1. I would like to know the melting point of this product

    1 answer
    1. The melting point of this product is not determined. However, Silicon Dioxide is reported to have a melting point of 1,610 °C or 2,930 °F. This information is published in the product Safety Data Sheet.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service