Skip to Content
Merck
All Photos(1)

Key Documents

26-0010

Sigma-Aldrich

Sand, white quartz

CP, beads

Synonym(s):

Quartz, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

CP

form

beads

availability

available only in Japan

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

René Schneider et al.
Optics express, 21(3), 3523-3539 (2013-03-14)
Illumination based on objective-type total internal reflection (TIR) is nowadays widely used in high-performance fluorescence microscopy. However, the desirable application of such setups for dark-field imaging of scattering entities is cumbersome due to the spatial overlap of illumination and detection
Sebastian Dochow et al.
Analytical and bioanalytical chemistry, 405(8), 2743-2746 (2013-02-02)
Three important technical innovations are reported here towards Raman-activated cell sorting. Firstly, a microfluidic chip made of quartz is introduced which integrates injection of single cells, trapping by laser fibres and sorting of cells. Secondly, a chip holder was designed
Lihui Feng et al.
Sensors (Basel, Switzerland), 13(9), 12482-12496 (2013-09-21)
A novel multi-fork z-axis gyroscope is presented in this paper. Different from traditional quartz gyroscopes, the lateral electrodes of the sense beam can be arranged in simple patterns; as a result, the fabrication is simplified. High sensitivity is achieved by
Xujia Jiang et al.
Environmental pollution (Barking, Essex : 1987), 174, 38-49 (2012-12-19)
The significance of biofilm on the transport and deposition behaviors of ZnO nanoparticles were examined under a series of environmentally relevant ionic strength at two fluid velocities of 4 m-d(-1) and 8 m-d(-1). Biofilm enhanced nanoparticles retention in porous media
Jenia Gutman et al.
Environmental science & technology, 47(1), 398-404 (2012-11-29)
This investigation is focused on the combined effect of bacterial physicochemical characteristics and motility on cell adhesion and deposition using a flow-through quartz crystal microbalance with dissipation (QCM-D). Three model flagellated strains with different degrees of motility were selected, including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service