Merck
  • Merit of quinacrine in the decrease of ingested sulfite-induced toxic action in rat brain.
Merit of quinacrine in the decrease of ingested sulfite-induced toxic action in rat brain.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-11-22)
Ceren Kencebay, Narin Derin, Ozlem Ozsoy, Dijle Kipmen-Korgun, Gamze Tanriover, Nihal Ozturk, Goksun Basaranlar, Piraye Yargicoglu-Akkiraz, Berna Sozen, Aysel Agar
ABSTRACT
We aimed at investigating the effects of sulfite-induced lipid peroxidation and apoptosis mediated by secretory phospholipase A2 (sPLA2) on somatosensory evoked potentials (SEP) alterations in rats. Thirty male albino Wistar rats were randomized into three experimental groups as follows; control (C), sodium metabisulfite treated (S), sodium metabisulfite+quinacrine treated (SQ). Sodium metabisulfite (100 mg/kg/day) was given by gastric gavage for 5 weeks and 10 mg/kg/day quinacrine was applied as a single dose of intraperitoneal injection for the same period. The latencies of SEP components were significantly prolonged in the S group and returned to control levels following quinacrine administration. Plasma-S-sulfonate level was increased in S and SQ groups. TBARS levels in the S group were significantly higher than those detected in controls. Quinacrine significantly decreased brain TBARS levels in the SQ group compared with the S group. Quinacrine treatment did not have an effect on the increased sPLA2 level of the sulfite administered group. Immunohistochemistry showed that sulfite caused an increase in caspase-3 and TUNEL positive cells, restored to control levels via quinacrine administration. This study showed that sPLA2 might play a role in ingested sulfite-induced SEP alterations, oxidative stress, apoptotic cell death and DNA damage in the brain.
Sigma-Aldrich
Sodium metabisulfite, JIS first grade, ≥95.0%
Sigma-Aldrich
Sodium metabisulfite, tested according to Ph. Eur.
Sigma-Aldrich
Quinacrine dihydrochloride, ≥90%
Sigma-Aldrich
Sodium metabisulfite, puriss. p.a., ACS reagent, reag. Ph. Eur., dry, 98-100.5%
Sigma-Aldrich
Sodium metabisulfite, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium metabisulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Sigma-Aldrich
Sodium metabisulfite, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium metabisulfite, reagent grade, 97%
Supelco
Sodium metabisulfite, analytical standard