Skip to Content
Merck
All Photos(1)

Key Documents

28-4010

Sigma-Aldrich

Sodium metabisulfite

≥95.0%

Synonym(s):

Sodium disulfite, Sodium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O5
CAS Number:
Molecular Weight:
190.11
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥95.0%
form:
powder

Assay

≥95.0%

form

powder

availability

available only in Japan

pH

4.5 (20 °C, 50 g/L)

mp

>300 °C (lit.)

SMILES string

[Na+].[Na+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2Na.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

HRZFUMHJMZEROT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noemí de Luna et al.
Disease models & mechanisms, 8(5), 467-472 (2015-03-13)
McArdle disease, also termed 'glycogen storage disease type V', is a disorder of skeletal muscle carbohydrate metabolism caused by inherited deficiency of the muscle-specific isoform of glycogen phosphorylase (GP-MM). It is an autosomic recessive disorder that is caused by mutations
Ivana Mara da Costa Machado Matos Carvalho et al.
Chemosphere, 82(7), 1056-1061 (2010-11-09)
Sodium metabisulfite is used in marine shrimp harvesting to prevent the occurrence of black spots. Shrimps are soaked in a sodium metabisulfite solution in ice, which is disposed of in sewages that run into marine canals, creating an environmental hazard.
Rosie F Davies et al.
Clinics in dermatology, 29(3), 311-315 (2011-04-19)
Human skin is exposed to a large variety of cosmetic allergens. Most allergic contact dermatitis occurs after exposure to fragrance, preservatives, and hair dyes. Such reactions can often be occult. As a result, a high index of suspicion is needed
Gisela Palma-Orozco et al.
Phytochemistry, 72(1), 82-88 (2010-11-23)
While a long shelf life for fruit products is highly desired, enzymatic browning is the main cause of quality loss in fruits and is therefore a main problem for the food industry. In this study polyphenol oxidase (PPO), the main
I U Nieves et al.
Bioresource technology, 102(13), 6959-6965 (2011-05-03)
Microaeration (injecting air into the headspace) improved the fermentation of hemicellulose hydrolysates obtained from the phosphoric acid pretreatment of sugarcane bagasse at 170°C for 10 min. In addition, with 10% slurries of phosphoric acid pretreated bagasse (180°C, 10 min), air

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service