71932
Sodium metabisulfite
tested according to Ph. Eur.
Synonym(s):
Natrii metabisulfis, Sodium disulfite, Sodium pyrosulfite
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Na2S2O5
CAS Number:
Molecular Weight:
190.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
>97.5%
form:
powder
Recommended Products
Agency
USP/NF
tested according to Ph. Eur.
Quality Level
Assay
>97.5%
form
powder
mp
>300 °C (lit.)
SMILES string
[Na+].[Na+].[O-]S(=O)S([O-])(=O)=O
InChI
1S/2Na.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2
InChI key
HRZFUMHJMZEROT-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Supplementary Hazards
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Rosie F Davies et al.
Clinics in dermatology, 29(3), 311-315 (2011-04-19)
Human skin is exposed to a large variety of cosmetic allergens. Most allergic contact dermatitis occurs after exposure to fragrance, preservatives, and hair dyes. Such reactions can often be occult. As a result, a high index of suspicion is needed
Ivana Mara da Costa Machado Matos Carvalho et al.
Chemosphere, 82(7), 1056-1061 (2010-11-09)
Sodium metabisulfite is used in marine shrimp harvesting to prevent the occurrence of black spots. Shrimps are soaked in a sodium metabisulfite solution in ice, which is disposed of in sewages that run into marine canals, creating an environmental hazard.
Jeffrey N Hemenway et al.
Journal of pharmaceutical sciences, 101(9), 3305-3318 (2012-05-23)
A 2,4-dinitrophenylhydrazine (DNPH) precolumn derivatization high-performance liquid chromatography-ultraviolet detection (HPLC-UV) method was developed to quantify levels of formaldehyde and acetaldehyde in polyethylene glycol (PEG) solutions. Formic acid and acetic acid were quantified by HPLC-UV. Samples of neat and aqueous PEG
I U Nieves et al.
Bioresource technology, 102(13), 6959-6965 (2011-05-03)
Microaeration (injecting air into the headspace) improved the fermentation of hemicellulose hydrolysates obtained from the phosphoric acid pretreatment of sugarcane bagasse at 170°C for 10 min. In addition, with 10% slurries of phosphoric acid pretreated bagasse (180°C, 10 min), air
Gisela Palma-Orozco et al.
Phytochemistry, 72(1), 82-88 (2010-11-23)
While a long shelf life for fruit products is highly desired, enzymatic browning is the main cause of quality loss in fruits and is therefore a main problem for the food industry. In this study polyphenol oxidase (PPO), the main
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service