Merck
  • Ligand contributions to the electronic structures of the oxidized cobalt(II) salen complexes.
Ligand contributions to the electronic structures of the oxidized cobalt(II) salen complexes.
Inorganic chemistry (2012-09-28)
Amélie Kochem, Hussein Kanso, Benoit Baptiste, Himanshu Arora, Christian Philouze, Olivier Jarjayes, Hervé Vezin, Dominique Luneau, Maylis Orio, Fabrice Thomas
ABSTRACT
Square planar cobalt(II) complexes of salen ligands N,N'-bis(3-tert-butyl-5R-salicylidene)-1,2-cyclohexanediamine), where R = OMe (1) and tert-butyl (2), were prepared. 1 and 2 were electrochemically reversibly oxidized into cations [1-H(2)O](+) and [2-H(2)O](+) in CH(2)Cl(2). The chemically generated [1-H(2)O](SbF(6))·0.68 H(2)O·0.82CH(2)Cl(2) and [2-H(2)O](SbF(6))·0.3H(2)O·0.85CH(2)Cl(2) were characterized by X-ray diffraction and NIR spectroscopy. Both complexes are paramagnetic species containing a square pyramidal cobalt ion coordinated at the apical position by an exogenous water molecule. They exhibit remarkable NIR bands at 1220 (7370 M(-1) cm(-1)) and 1060 nm (5560 M(-1) cm(-1)), respectively, assigned to a CT transition. DFT calculations and magnetic measurements confirm the paramagnetic (S = 1) ground spin state of the cations. They show that more than 70% of the total spin density in [1-H(2)O](+) and [2-H(2)O](+) is localized on the metal, the remaining spin density being distributed over the aromatic rings (30% phenoxyl character). In the presence of N-methylimidazole 1 and 2 are irreversibly oxidized by air into the genuine octahedral cobalt(III) bis(phenolate) complexes [1-im(2)](+) and [2-im(2)](+), the former being structurally characterized. Neither [1-im(2)](+) nor [2-im(2)](+) exhibits a NIR feature in its electronic spectrum. 1 and 2 were electrochemically two-electron oxidized into [1](2+) and [2](2+). The cations were identified as Co(III)-phenoxyl species by their characteristic absorption band at ca. 400 nm in the UV-vis spectrum. Coordination of the phenoxyl radical to the cobalt(III) metal ion is evidenced by the EPR signal centered at g = 2.00.
Product Description
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
N,N′-Bis(salicylidene)ethylenediamine, 98%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer