07-2180
Dichloromethane
JIS special grade, ≥99.0%
Synonym(s):
Methylene chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
grade:
JIS special grade
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)
6.83 psi ( 20 °C)
Recommended Products
grade
JIS special grade
vapor density
2.9 (vs air)
vapor pressure
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)
Assay
≥99.0%
form
liquid
autoignition temp.
1223 °F
expl. lim.
22 %
availability
available only in Japan
dilution
(for analytical testing)
refractive index
n20/D 1.424 (lit.)
bp
39.8-40 °C (lit.)
mp
−95 °C (lit.)
density
1.325 g/mL at 25 °C (lit.)
SMILES string
ClCCl
InChI
1S/CH2Cl2/c2-1-3/h1H2
InChI key
YMWUJEATGCHHMB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
WGK
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Aria Nouri et al.
Spine, 40(14), 1092-1100 (2015-04-22)
Ambispective study. To determine whether MRI parameters improve the predictive performance of a validated clinical prediction rule used to assess functional outcomes in surgical patients with DCM. Degenerative cervical myelopathy (DCM) is the most common cause of spinal cord dysfunction
Damian M Herz et al.
Brain : a journal of neurology, 138(Pt 6), 1658-1666 (2015-04-18)
Dopaminergic signalling in the striatum contributes to reinforcement of actions and motivational enhancement of motor vigour. Parkinson's disease leads to progressive dopaminergic denervation of the striatum, impairing the function of cortico-basal ganglia networks. While levodopa therapy alleviates basal ganglia dysfunction
Molamma P Prabhakaran et al.
Materials science & engineering. C, Materials for biological applications, 56, 66-73 (2015-08-08)
Advanced engineering of materials for the development of drug delivery devices provides scope for novel and versatile strategies for treatment of various diseases. 'Electrospraying' was used to prepare PLGA microparticles and further encapsulate the drug, metronidazole (Met) within the particles
Angharad M Roberts et al.
Science translational medicine, 7(270), 270ra6-270ra6 (2015-01-16)
The recent discovery of heterozygous human mutations that truncate full-length titin (TTN, an abundant structural, sensory, and signaling filament in muscle) as a common cause of end-stage dilated cardiomyopathy (DCM) promises new prospects for improving heart failure management. However, realization
K E Mauck et al.
Integrative and comparative biology, 54(2), 193-209 (2014-04-22)
Recent research suggests that plant viruses, and other pathogens, frequently alter host-plant phenotypes in ways that facilitate transmission by arthropod vectors. However, many viruses infect multiple hosts, raising questions about whether these pathogens are capable of inducing transmission-facilitating phenotypes in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service