Skip to Content
Merck
All Photos(1)

Key Documents

236071

Sigma-Aldrich

N,N′-Bis(salicylidene)ethylenediamine

98%

Synonym(s):

2,2′-Ethylenebis(nitrilomethylidene)diphenol, N,N′-Ethylenebis(salicylimine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2N=CHC6H4OH)2
CAS Number:
Molecular Weight:
268.31
Beilstein:
535296
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

mp

127-128 °C (lit.)

solubility

water: insoluble(lit.)

functional group

amine
imine

SMILES string

Oc1ccccc1\C=N\CC\N=C\c2ccccc2O

InChI

1S/C16H16N2O2/c19-15-7-3-1-5-13(15)11-17-9-10-18-12-14-6-2-4-8-16(14)20/h1-8,11-12,19-20H,9-10H2/b17-11+,18-12+

InChI key

VEUMANXWQDHAJV-JYFOCSDGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N′-Bis(salicylidene)ethylenediamine (salen) reacts with ferric chloride in acetone solution to yield the complex (Fe salen Cl)[1].

Application

N,N′-Bis(salicylidene)ethylenediamine (salen) immobilized on surfactant-coated alumina has been used as complexing agent for on-line preconcentration of copper and lead by flow injection-flame atomic absorption spectrometry (FI-AAS)[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydration of epoxides on [CoIII salen)] encapsulated in silica-based nanoreactors.
Bo Li et al.
Angewandte Chemie (International ed. in English), 51(46), 11517-11521 (2012-10-16)
Andrea Pappalardo et al.
The Journal of organic chemistry, 77(17), 7684-7687 (2012-08-16)
A pair of diastereomeric salen cavitands and their uranyl complexes combine a chiral (R,R) salen bridge and an inherent chiral tris-bridged quinoxaline cup within the same molecule. Whereas the free ligands show a preference for the same enantiomer of an
Mohammad Azam et al.
Dalton transactions (Cambridge, England : 2003), 41(35), 10854-10864 (2012-08-04)
We have reported the synthesis of a novel salen ligand and its mononuclear Pd-salen complexes derived from 2-{[2-hydroxy-3-{[(E)-(2-hydroxyphenyl)methylidene]amino}propyl)imino]methyl}phenol. The newly synthesized and isolated Pd(II) complexes have been identified and fully characterized by various physico-chemical studies viz., elemental analyses, IR, UV-Vis
Lakshmi Kanta Das et al.
Dalton transactions (Cambridge, England : 2003), 41(36), 11009-11017 (2012-08-04)
Two new trinuclear hetero-metallic copper(II)-zinc(II) complexes [(CuL)(2)Zn(N(3))(2)] (1A and 1B) have been synthesized using [CuL] as a so-called "metalloligand" (where H(2)L = N,N'-bis(salicylidene)-1,3-propanediamine) and structurally characterized. Complexes 1A and 1B have the same molecular formula but crystallize in different crystal
Erin E Podlesny et al.
Organic letters, 14(18), 4862-4865 (2012-09-04)
The selective oxidation of a series of functionalized 8,8'-hydroxylated binaphthols to binaphtho-para- and binaphtho-ortho-quinones has been realized using either a Co-salen catalyst or ortho-iodoxybenzoic acid. A unique spirocyclic bis-spironaphthalenone was also obtained in good yield via a phenyliodonium diacetate promoted

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service