Merck
  • Synthesis and characterization of polyacetylene with side-chain thiophene functionality.
Synthesis and characterization of polyacetylene with side-chain thiophene functionality.
International journal of molecular sciences (2009-03-28)
Banu Koz, Baris Kiskan, Yusuf Yagci
ABSTRACT
A new polyacetylene derivative with electroactive thiophene substituent, namely poly(2-methylbut-2-enyl thiophene-3-carboxylate) was synthesized and characterized. For this purpose, novel acetylene monomer was synthesized by the reaction of 3-thiophenecarboxylic acid with propargyl bromide and polymerized with a Rh catalyst to give the corresponding polymer. The chemical structure of the polymer was characterized to comprise the conjugated backbone and electroactive thiophene side group. UV spectral changes of the polymer with temperature were also studied. The polymer exhibited better thermal stability than the unsubstituted polyacetylenes.
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Supelco
Toluene, analytical standard
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
3-Thiophenecarboxylic acid, 99%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, JIS 1000, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%