Skip to Content
Merck
All Photos(1)

Key Documents

648566

Sigma-Aldrich

Toluene

suitable for HPLC, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

HPLC grade

Quality Level

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

technique(s)

HPLC: suitable

impurities

≤0.02% water

evapn. residue

≤0.0005%

color

colorless

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

UV absorption

λ: 290 nm Amax: 0.30
λ: 310 nm Amax: 0.05
λ: 350 nm Amax: 0.004

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluene, a benzene analog, is widely employed as an organic solvent. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations).[1] Its anaerobic biodegradation to CO2, by the denitrifying bacterium Thauera arornatica has been reported.[2] It forms syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.[3]

Application

Toluene may be used in the following studies:
  • As a doping agent during the quantitative estimation of triacylglycerols (TAGs) by liquid chromatography-atmospheric pressure photoionization-mass spectrometry (LC-APPIMS). It has been reported to improve the sensitivity of analysis.[4]
  • For the decontamination of microbial cultures and lyse bacterial cells in bacterial enzyme assays.[1]
  • As a solvent for the preparation of 4-arm polylactide-based (PLA) star oligomer.[5]

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sheng-Suan Cai et al.
Journal of chromatography. A, 1173(1-2), 88-97 (2007-10-30)
In this work, we evaluate the performance of liquid chromatography-atmospheric pressure photoionization-mass spectrometry (LC-APPI-MS) for non-aqueous reversed phase analysis of six triacylglycerol model compounds using six binary mobile phases including MeOH/iPrOH, MeOH/CHCl(3), MeOH/CH(2)Cl(2), CH(3)CN/iPrOH, CH(3)CN/CHCl(3), and CH(3)CN/CH(2)Cl(2). All mobile phases
T Biegert et al.
European journal of biochemistry, 238(3), 661-668 (1996-06-15)
Toluene is degraded anoxically to CO2 by the denitrifying bacterium Thauera aromatica. Toluene first becomes oxidized to benzoyl-CoA by O2-independent reactions. Benzoyl-CoA is then reduced to non-aromatic products by benzoyl-CoA reductase. We set out to study the reactions employed for
Structural study on syndiotactic polystyrene: 2. Crystal structure of molecular compound with toluene.
Chatani Y, et al.
Polymer, 34(8), 1620-1620 (1993)
A Pseudomonas thrives in high concentrations of toluene.
Inoue A and Horikoshi K.
Nature, 338, 264-266 (1989)
V Melissinaki et al.
Biofabrication, 3(4), 045005-045005 (2011-09-21)
This study reports on the production of high-resolution 3D structures of polylactide-based materials via multi-photon polymerization and explores their use as neural tissue engineering scaffolds. To achieve this, a liquid polylactide resin was synthesized in house and rendered photocurable via

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service