Skip to Content
Merck
All Photos(2)

Key Documents

34866

Sigma-Aldrich

Toluene

suitable for HPLC, 99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
41116105
eCl@ss:
39011102
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
UV/Vis spectroscopy: suitable
application(s):
food and beverages
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

grade

HPLC grade

Quality Level

Agency

suitable for EPA 1613

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

99.9%

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

technique(s)

HPLC: suitable
UV/Vis spectroscopy: suitable

impurities

≤0.0005% non-volatile matter
≤0.0005% thiophene
≤0.001% free acid (as HCl)
≤0.02% water (Karl Fischer)

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

UV absorption

λ: 286 nm Amax: ≤1.00
λ: 288 nm Amax: ≤0.50
λ: 293 nm Amax: ≤0.20
λ: 300 nm Amax: ≤0.10
λ: 310 nm Amax: ≤0.05
λ: 335 nm Amax: ≤0.02
λ: 350 nm Amax: ≤0.004

application(s)

food and beverages

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluene, a benzene analog, is widely employed as an organic solvent. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations). Its anaerobic biodegradation to CO2, by the denitrifying bacterium Thauera arornatica has been reported. It forms a syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.

Application

Toluene may be used as a doping agent during the quantitative estimation of triacylglycerols (TAGs) by liquid chromatography-atmospheric pressure photoionization-mass spectrometry (LC-APPIMS). It has been reported to improve the sensitivity of analysis. It may be used as solvent for the preparation of 4-arm polylactide-based (PLA) star oligomer.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Other Notes

Important notice
  • The article number 34866-4X2.5L will be discontinued. Please order the single bottle 34866-2.5L which is physically identical with the same exact specifications.
  • The article number 34866-6X1L will be discontinued. Please order the single bottle 34866-1L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Direct laser writing of 3D scaffolds for neural tissue engineering applications.
Melissinaki V, et al.
Biofabrication, 3(4), Direct laser writing-Direct laser writing (2011)
Sheng-Suan Cai et al.
Journal of chromatography. A, 1173(1-2), 88-97 (2007-10-30)
In this work, we evaluate the performance of liquid chromatography-atmospheric pressure photoionization-mass spectrometry (LC-APPI-MS) for non-aqueous reversed phase analysis of six triacylglycerol model compounds using six binary mobile phases including MeOH/iPrOH, MeOH/CHCl(3), MeOH/CH(2)Cl(2), CH(3)CN/iPrOH, CH(3)CN/CHCl(3), and CH(3)CN/CH(2)Cl(2). All mobile phases
T Biegert et al.
European journal of biochemistry, 238(3), 661-668 (1996-06-15)
Toluene is degraded anoxically to CO2 by the denitrifying bacterium Thauera aromatica. Toluene first becomes oxidized to benzoyl-CoA by O2-independent reactions. Benzoyl-CoA is then reduced to non-aromatic products by benzoyl-CoA reductase. We set out to study the reactions employed for
A Pseudomonas thrives in high concentrations of toluene.
Inoue A and Horikoshi K.
Nature, 338, 264-266 (1989)
Structural study on syndiotactic polystyrene: 2. Crystal structure of molecular compound with toluene.
Chatani Y, et al.
Polymer, 34(8), 1620-1620 (1993)

Articles

Nonylphenols (NP) are ubiquitous substances that have been detected in highly diverse foodstuff. As breakdown products of nonylphenol ethoxylates, NP could end up in the aquatic environment. This article presents a rapid method for NP detection in surface water by planar solid phase extraction – planar yeast estrogen screen (pSPE-pYES).

Nonylphenols (NP) are ubiquitous substances that have been detected in highly diverse foodstuff. As breakdown products of nonylphenol ethoxylates, NP could end up in the aquatic environment. This article presents a rapid method for NP detection in surface water by planar solid phase extraction – planar yeast estrogen screen (pSPE-pYES).

Nonylphenols (NP) are ubiquitous substances that have been detected in highly diverse foodstuff. As breakdown products of nonylphenol ethoxylates, NP could end up in the aquatic environment. This article presents a rapid method for NP detection in surface water by planar solid phase extraction – planar yeast estrogen screen (pSPE-pYES).

Nonylphenols (NP) are ubiquitous substances that have been detected in highly diverse foodstuff. As breakdown products of nonylphenol ethoxylates, NP could end up in the aquatic environment. This article presents a rapid method for NP detection in surface water by planar solid phase extraction – planar yeast estrogen screen (pSPE-pYES).

See All

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service