Merck
  • HNF4A and GATA6 Loss Reveals Therapeutically Actionable Subtypes in Pancreatic Cancer.
HNF4A and GATA6 Loss Reveals Therapeutically Actionable Subtypes in Pancreatic Cancer.
Cell reports (2020-05-14)
Holly Brunton, Giuseppina Caligiuri, Richard Cunningham, Rosie Upstill-Goddard, Ulla-Maja Bailey, Ian M Garner, Craig Nourse, Stephan Dreyer, Marc Jones, Kim Moran-Jones, Derek W Wright, Viola Paulus-Hock, Colin Nixon, Gemma Thomson, Nigel B Jamieson, Grant A McGregor, Lisa Evers, Colin J McKay, Aditi Gulati, Rachel Brough, Ilirjana Bajrami, Stephen J Pettitt, Michele L Dziubinski, Simon T Barry, Robert Grützmann, Robert Brown, Edward Curry, Marina Pajic, Elizabeth A Musgrove, Gloria M Petersen, Emma Shanks, Alan Ashworth, Howard C Crawford, Diane M Simeone, Fieke E M Froeling, Christopher J Lord, Debabrata Mukhopadhyay, Christian Pilarsky, Sean E Grimmond, Jennifer P Morton, Owen J Sansom, David K Chang, Peter J Bailey, Andrew V Biankin
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) can be divided into transcriptomic subtypes with two broad lineages referred to as classical (pancreatic) and squamous. We find that these two subtypes are driven by distinct metabolic phenotypes. Loss of genes that drive endodermal lineage specification, HNF4A and GATA6, switch metabolic profiles from classical (pancreatic) to predominantly squamous, with glycogen synthase kinase 3 beta (GSK3β) a key regulator of glycolysis. Pharmacological inhibition of GSK3β results in selective sensitivity in the squamous subtype; however, a subset of these squamous patient-derived cell lines (PDCLs) acquires rapid drug tolerance. Using chromatin accessibility maps, we demonstrate that the squamous subtype can be further classified using chromatin accessibility to predict responsiveness and tolerance to GSK3β inhibitors. Our findings demonstrate that distinct patterns of chromatin accessibility can be used to identify patient subgroups that are indistinguishable by gene expression profiles, highlighting the utility of chromatin-based biomarkers for patient selection in the treatment of PDAC.
Sigma-Aldrich
6-Aminonicotinamide, 99%
Sigma-Aldrich
apo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
O-Phosphorylethanolamine
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
MISSION® esiRNA, targeting human HNF4A
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Millipore
Millex PVDF syringe filter, pore size 0.45 μm, diam. 4 mm, sterile, hydrophilic
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Carnitine hydrochloride, synthetic, ≥98%
Sigma-Aldrich
MISSION® esiRNA, targeting human PLK1
Sigma-Aldrich
MISSION® esiRNA, targeting human GATA6
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Oligomycin from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC)
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder