Skip to Content
Merck
All Photos(3)

Key Documents

E1905

Sigma-Aldrich

(+)-Etomoxir sodium salt hydrate

≥98% (HPLC), powder, CPT-1 inhibitor

Synonym(s):

R(+)-2-[6-(4-Chlorophenoxy)hexyl]-oxirane-2-carboxylic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18ClO4 · Na · H2O
CAS Number:
Molecular Weight:
338.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

H2O: freely soluble

storage temp.

2-8°C

SMILES string

O.[Na+].[O-]C(=O)[C@@]2(CCCCCCOc1ccc(Cl)cc1)CO2

InChI

1S/C15H19ClO4.Na.H2O/c16-12-5-7-13(8-6-12)19-10-4-2-1-3-9-15(11-20-15)14(17)18;;/h5-8H,1-4,9-11H2,(H,17,18);;1H2/q;+1;/p-1/t15-;;/m1../s1

InChI key

WENJLJJSJDAJDN-QCUBGVIVSA-M

General description

(+)-Etomoxir sodium salt hydrate belongs to oxirane carboxylic acid group of compounds and mediates metabolic channeling of fatty acid precursors.[1] It favors oxidative stress in T cells[2] and prevents T cell differentiation.[3] It inhibits fatty acid oxidation and promotes hunger and food intake.[4] Etomoxir impairs myeloid-derived suppressor cells mediated tumor and could have potential therapeutic potential.[5]

Application

(+)-Etomoxir sodium salt hydrate has been used as an inhibitor of carnitine palmitoyltransferase 1 (CPT-1) in breast tumor cell lines[6] and mice tumor.[7] It has also been used as an inhibitor of fatty acid oxidation in human primary prostate epithelial cells[8] and lymphocytes.[9]

Biochem/physiol Actions

Irreversible O-carnitine palmitoyltransferase-1 (CPT-1) inhibitor; PPARα activator

Packaging

Store tightly sealed, protected from exposure to light

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The CPT1a inhibitor, etomoxir induces severe oxidative stress at commonly used concentrations
OConnor RS, et al.
Scientific Reports, 8(1), 6289-6289 (2018)
Etomoxir mediates differential metabolic channeling of fatty acid and glycerol precursors into cardiolipin in H9c2 cells
Xu FY, et al.
Journal of Lipid Research, 44(2), 415-423 (2003)
L-carnitine ameliorates the liver inflammatory response by regulating carnitine palmitoyltransferase I-dependent PPAR gamma signaling
Jiang F, et al.
Molecular Medicine Reports, 13(2), 1320-1328 (2016)
Ibrutinib Resistance Is Reduced By Fatty Acid Oxidation Inhibition In Primary CLL Lymphocytes
Galicia-Vazquez G and Aloyz R
Frontiers in Oncology, 8, 411-411 (2018)
Organoids model distinct Vitamin E effects at different stages of prostate cancer evolution
Njoroge RN, et al.
Scientific Reports, 7(1), 16285-16285 (2017)

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

  1. What are the specific inhibitor recommendations for beta oxidation?

    1 answer
    1. For an inhibitor for beta oxidation, the following products may be of interest:
      •E1905 (+)-Etomoxir sodium salt hydrate
      •5.09455 InSolution Etomoxir
      •M5199 Meldonium dihydrate

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service