P0503
O-Phosphorylethanolamine
Synonym(s):
2-Aminoethyl dihydrogen phosphate, O-Phosphocolamine, O-Phosphoethanolamine, Colaminephosphoric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH2CH2CH2OPO3H2
CAS Number:
Molecular Weight:
141.06
Beilstein:
1758916
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
form
powder
Quality Level
storage temp.
−20°C
SMILES string
NCCOP(O)(O)=O
InChI
1S/C2H8NO4P/c3-1-2-7-8(4,5)6/h1-3H2,(H2,4,5,6)
InChI key
SUHOOTKUPISOBE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
O-Phosphorylethanolamine has been used:
- in LHC-8 medium as a supplement in culturing the transformed human liver epithelial (THLE)-2 and THLE-3 cells
- in C-reactive protein (CRP) binding sucrose flotation experiment
- to incubate phosphomonoesters with anion-exchange chromatography (AEC) fractions for the detection of phosphatase activity
- to add in the washed beads for the preparation of phosphorylethanolamine (Pet) -conjugated sepharose column for chromatographic purification of mutant CRP
Biochem/physiol Actions
Phosphorylethanolamine participates in phospholipid metabolism. Its release can be stimulated occasionally by depolarizing stimuli. Reduction of phosphorylethanolamine levels has been observed in Alzheimer′s and Huntington′s disease.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The phosphocholine-binding pocket on C-reactive protein is necessary for initial protection of mice against pneumococcal infection
Gang TB, et al.
Test, jbc-M112 (2012)
Phosphoethanolamine-complexed C-reactive protein: a pharmacological-like macromolecule that binds to native low-density lipoprotein in human serum
Singh SK, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 394(1-2), 94-98 (2008)
Phosphoethanolamine and ethanolamine are decreased in Alzheimer's disease and Huntington's disease
Ellison David W, et al.
Brain Research, 417(2), 389-392 (1987)
Lin Zhu et al.
ACS nano, 6(4), 3491-3498 (2012-03-14)
A novel "smart" multifunctional drug delivery system was successfully developed to respond to the up-regulated matrix metalloprotease 2 (MMP2) in the tumor microenvironment and improve cancer cell-specific delivery of loaded drugs. The system represents a surface-functionalized liposomal nanocarrier, for which
Christopher Wanty et al.
Journal of molecular biology, 425(18), 3389-3402 (2013-07-03)
Gram-negative bacteria possess an outer membrane envelope consisting of an outer leaflet of lipopolysaccharides, also called endotoxins, which protect the pathogen from antimicrobial peptides and have multifaceted roles in virulence. Lipopolysaccharide consists of a glycan moiety attached to lipid A
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service