Skip to Content
Merck
All Photos(5)

Key Documents

C0283

Sigma-Aldrich

L-Carnitine hydrochloride

≥98%, synthetic

Synonym(s):

(−)-β-Hydroxy-γ-(trimethylammonio)butyrate, Vitamin BT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2CH(OH)CH2CO2H · Cl-
CAS Number:
Molecular Weight:
197.66
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.79

Product Name

L-Carnitine hydrochloride, synthetic, ≥98%

biological source

synthetic

Assay

≥98%

form

powder

color

white

mp

142 °C

application(s)

cell analysis

SMILES string

[Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O

InChI

1S/C7H15NO3.ClH/c1-8(2,3)5-6(9)4-7(10)11;/h6,9H,4-5H2,1-3H3;1H/t6-;/m1./s1

InChI key

JXXCENBLGFBQJM-FYZOBXCZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Carnitine hydrochloride has been used to study ergothioneine transporter SLC22A4 and carnitine transporter SLC22A5. It has also been used to allow the entry of palmitic acid into the mitochondria.

Biochem/physiol Actions

Carnitine is a quaternary amine that occurs naturally in most mammalian tissue. It is present in relatively high concentrations in skeletal muscle and heart where it is involved in regulating energy metabolism. It shifts glucose metabolism from glycolysis to glycogen storage and enhances the transport of long chain fatty acids into the mitochondria where they are oxidized for energy production.
Carnitine is biosynthesized from lysine. It might be associated with the energy production from branched chain amino acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Isabelle Lelong-Rebel et al.
Anticancer research, 28(1A), 55-68 (2008-04-04)
Two chemosensitive cell lines, LoVo-fusoid (LoVo-f) and LoVo-small cells (LoVo-sc) were derived from the original LoVo cell line. These two variants and the multidrug-resistant (MDR) cell line LoVo-Dox were screened for various properties. In non-permeabilized cells, only LoVo-sc showed mucin-2
Substrate discrimination by ergothioneine transporter SLC22A4 and carnitine transporter SLC22A5: Gain-of-function by interchange of selected amino acids
Petra Bacher.
Biochimica et Biophysica Acta, 1788(12), 2594-2602 (2009)
Giulia Mollica et al.
Digestive and liver disease : official journal of the Italian Society of Gastroenterology and the Italian Association for the Study of the Liver, 52(3), 314-323 (2019-10-15)
Non-alcoholic fatty liver disease (NAFLD) is a common cause of chronic liver disorder. NAFLD, associated lipotoxicity, fibrosis, oxidative stress, and altered mitochondrial metabolism, is responsible for systemic inflammation, which contributes to organ dysfunction in extrahepatic tissues, including the heart. We
R E L Cabral et al.
Andrology, 6(1), 236-246 (2017-10-04)
Doxorubicin has been largely used in anticancer therapy in adults, adolescents, and children. The efficacy of l-carnitine as an antioxidant substance has been confirmed both in humans and rats. Carnitine, present in testis and epididymis, is involved in sperm maturation.
K Kasperek et al.
Polish journal of veterinary sciences, 21(4), 811-813 (2019-01-04)
The aim of our study was to determine the influence of L-carnitine (L-CAR) on the cellular parameters of hen erythrocytes during a 48 hour exposure to L-CAR at concentrations of 25, 50 and 100 µg/mL in nutrient-deficient medium. Cell morphology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service